Platinum Crucibles
Prohibit contamination at high temperatures
  • Oxidation and corrosion resistance
  • Easy to clean
These crucibles offer high temperature strength up to 2192°F (1200°C) and high melting point. Platinum offers oxidation resistance in air and oxygen; and corrosion resistance in acids and melted salts.
Protect your investment with platinum tongs that reduce possible damage and scratches to platinum crucible when hot.
Manufactured in standard form. Lids and tongs sold separately.
303,000.00 - ₹484,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Inside Rim Diameter (mm) Material
Availability Pricing
Platinum Crucible, standard form, 10 mL, 1/ea WW-17830-15 Mfr # GCS10
COMPARE
10 25 Platinum
₹303,000.00INR / Each
Platinum Crucible, standard form, 15 mL, 1/ea WW-17830-25 Mfr # GCS15
COMPARE
15 30 Platinum
₹341,000.00INR / Each
Platinum Crucible, standard form, 25 mL, 1/ea WW-17830-45 Mfr # GCS25
COMPARE
25 36 Platinum
Call For Price
Platinum Crucible, standard form, 30 mL, 1/ea WW-17830-55 Mfr # GCS30
COMPARE
30 40 Platinum
Call For Price
Platinum Crucible, standard form, 40 mL, 1/ea WW-17830-65 Mfr # GCS40
COMPARE
40 42 Platinum
Call For Price
Platinum Crucible, standard form, 50 mL, 1/ea WW-17830-75 Mfr # GCS50
COMPARE
50 45 Platinum
Call For Price
Platinum Crucible, wide form, 50 mL, 1/ea WW-17830-82 Mfr # GD50
COMPARE
50 53 Platinum
₹484,000.00INR / Each
Platinum Crucible, wide form, 75 mL, 1/ea WW-17830-86 Mfr # GD75
COMPARE
75 65 Platinum
Call For Price
Platinum Crucible, wide form, 100 mL, 1/ea WW-17830-88 Mfr # GD100
COMPARE
100 75 Platinum
Call For Price
Showing 1- 9 of 9