PharmaFluor® Reducer Elbow Fittings, PFA
High-purity compression fittings designed for critical fluid and gas applications
  • Ultra-clean PFA material construction
  • Excellent chemical resistance
  • Tool-free installation for faster setups
  • Reliable performance in critical systems
PharmaFluor® PFA compression fittings are engineered to deliver reliable, contamination-free performance in critical fluid and gas handling systems. Manufactured from 100% virgin high-purity PFA, these fittings ensure exceptional chemical resistance and ultra-clean operation, making them ideal for applications in biopharmaceutical, semiconductor, chemical, and food processing environments. These fittings do not require any tools, enabling fast, efficient installation while maintaining secure, leak-resistant connections.
16,900.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Compression Size / Tube Od (Port 2) (in) Flow Pattern
Availability Pricing
PharmaFluor Compression Fitting, Elbow Reducer, PFA, 3/4" OD x 1/2" OD 5014468 Mfr # G1077384
COMPARE
1/2 3/4 Elbow
Call For Price
PharmaFluor Compression Fitting, Elbow Reducer, PFA, 1/2" OD x 3/8" OD 5014469 Mfr # G1077390
COMPARE
3/8 1/2 Elbow
₹16,900.00INR / Each