PharmaFluor® Ferrule Fittings, PFA
High-purity compression fittings designed for critical fluid and gas applications
  • Ultra-clean PFA material construction
  • Excellent chemical resistance
  • Tool-free installation for faster setups
  • Reliable performance in critical systems
PharmaFluor® PFA compression fittings are engineered to deliver reliable, contamination-free performance in critical fluid and gas handling systems. Manufactured from 100% virgin high-purity PFA, these fittings ensure exceptional chemical resistance and ultra-clean operation, making them ideal for applications in biopharmaceutical, semiconductor, chemical, and food processing environments. These fittings do not require any tools, enabling fast, efficient installation while maintaining secure, leak-resistant connections.
1,400.00 - ₹2,100.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Connection Type
Availability Pricing
PharmaFluor Ferrule Fitting, PFA, 1/8" OD 5014474 Mfr # G1077073
COMPARE
1/8 Ferrule
₹1,400.00INR / Each
PharmaFluor Ferrule Fitting, PFA, 1/2" OD 5014475 Mfr # G1077035
COMPARE
1/2 Ferrule
₹1,800.00INR / Each
PharmaFluor Ferrule Fitting, PFA, 1/4" OD 5014476 Mfr # G1077036
COMPARE
1/4 Ferrule
₹1,500.00INR / Each
PharmaFluor Ferrule Fitting, PFA, 3/4" OD 5014477 Mfr # G1077037
COMPARE
3/4 Ferrule
₹2,100.00INR / Each
PharmaFluor Ferrule Fitting, PFA, 3/8" OD 5014478 Mfr # G1077038
COMPARE
3/8 Ferrule
₹1,700.00INR / Each