PharmaFluor Ferrule Fitting, PFA, 3/4" OD
Representative image only.
PharmaFluor Ferrule Fitting, PFA, 3/4" OD
PharmaFluor | Mfr: G1077037| Item: 5014477
High-purity compression fittings designed for critical fluid and gas applications
₹2,100.00INR / Each
₹2,100.00INR / Each
Product Description
Key Features
  • Ultra-clean PFA material construction
  • Excellent chemical resistance
  • Tool-free installation for faster setups
  • Reliable performance in critical systems
More About this Item
PharmaFluor® PFA compression fittings are engineered to deliver reliable, contamination-free performance in critical fluid and gas handling systems. Manufactured from 100% virgin high-purity PFA, these fittings ensure exceptional chemical resistance and ultra-clean operation, making them ideal for applications in biopharmaceutical, semiconductor, chemical, and food processing environments. These fittings do not require any tools, enabling fast, efficient installation while maintaining secure, leak-resistant connections.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPFA
  • Chemical ResistanceChemically inert; corrosion resistant
  • Number Of Ports1
  • Connection TypeFerrule
  • Compression Size / Tube Od (Port 1) (in)3/4
  • Compression Size / Tube Od (Port 1) (mm)19.05
  • Max Temperature (° F)212
  • Max Temperature (° C)100
  • Quantity1
  • DescriptionPharmaFluor® Ferrule Fitting, PFA, 3/4" OD
  • Warranty12 months from date of first sale