Peak Scientific Precision Air Compressors
A dedicated, fully integrated compressed air source for Precision gas generators
  • Compressed air supply for Precision Nitrogen and Zero Air generators
  • Single unit can supply both Precision Nitrogen and Zero Air generators in a stack
  • Duplex compressor mounting system to dampen noise and vibration
Precision Air Compressor is designed specifically to provide a dedicated, fully integrated compressed air source for Precision Nitrogen and Precision Zero Air gas generators. Compressor can be added to any Precision stack, typically at the bottom, to give you a complete self-contained GC gas solution, even when no house supply is available. Its duplex compressor mounting system features low noise and vibration for quiet operation.
3,392,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (bar) Power (VAC) Description
Availability Pricing
PEAK Scientific Precision Air Compressor; 230 VAC 8644339 Mfr # 65-0555
COMPARE
10 230 Air Compressor; 230 VAC
₹3,392,000.00INR / Each
PEAK Scientific Precision Air Compressor; 110 VAC 8644340 Mfr # 65-1555
COMPARE
10 110 Air Compressor; 110 VAC
₹3,392,000.00INR / Each