Open-Walled Containers

Handles provide easy lifting
Durable, corrosion-resistant HDPE containers have mesh sides and bottom for easy viewing, rinsing, and drying. Straight sides allow stable stacking- all four sizes stack together.
5,700.00 - ₹10,500.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Length (in) Width (in)
Availability Pricing
Akro-Mils 37-208 Mesh-Walled Storage Container, HDPE, Stackable, 16 x 12 x 8" 4753000 Mfr # 37-208
COMPARE
8 16 12
₹5,700.00INR / Each
Akro-Mils 37-608 Mesh-Walled Storage Container, HDPE, Stackable, 24 x 16 x 8" 4753005 Mfr # 37-608
COMPARE
8 24 16
₹8,400.00INR / Each
Akro-Mils 37-612 Mesh-Walled Storage Container, HDPE, Stackable, 24 x 16 x 12" 4753010 Mfr # 37-612
COMPARE
12 24 16
₹9,000.00INR / Each
Akro-Mils 37-616 Mesh-Walled Storage Container, HDPE, Stackable, 24 x 16 x 16" 4753015 Mfr # 37-616
COMPARE
16 24 16
₹10,500.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat