Ohaus® YA Handheld Balances
Clamshell case design protects scale during transport and storage
  • Rugged ABS construction with sealed front panel
  • Stainless steel weighing pan
  • Unique hinged cover
  • Energy saving auto shut-off
  • Built-in stability and over/under load indicators
  • Bright, backlit LCD
31,200.00 - ₹35,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Capacity (g) Readability (g)
Availability Pricing
Ohaus YA102 YA Handheld Balance, 100 x 0.01g (High Contrast Lcd) 1111007 Mfr # YA102
COMPARE
2 AAA (included) 100 0.01
₹35,700.00INR / Each
Ohaus YA302 YA Handheld Balance, 300 x 0.05G (High Contrast Lcd) 1111008 Mfr # YA302
COMPARE
2 AAA (included) 300 0.05
₹31,200.00INR / Each