Ohaus® Valor 3000 Xtreme Portion Control Washdown Scales
Extreme performance in an extremely rugged stainless steel design
  • Fast 2-second stabilization time
  • Multiple weighing units include g, fractional or decimal oz, kg, lb, lb:oz
  • All stainless steel design with shock and overload protection for tough environments
  • Washdown IP65-rated models may be rinsed clean with sprayed water
These compact industrial balances offer an extremely rugged design for long-term performance in tough environments. Valor is NSF certified, USDA-AMS accepted, and supports HACCP-certified systems. Wash down IP65-rated models may be rinsed clean with sprayed water. No exposed screws or edges make these balances perfect for food testing and service environments and offer easy clean-up. Ohaus’ classic SmarText™ driven interface includes a brilliant backlit display and Yes/No menu navigation offers fast, user-intuitive operation — adjustable stability settings and resolutions allow the Valor 3000 series to operate accurately in drafty and unstable environments.
185,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Ohaus V31XW301 Valor 3000 xtremePortion Control Washdown Scale, 300g x 0.2g; Ip-65/Nema 1101185 Mfr # V31XW301
COMPARE
0.3 0.66 115
₹185,000.00INR / Each
Ohaus V31XW3 Valor 3000 Xtreme Portion Control Washdown Scale, 3000g x 1g; Ip-65/Nema 1101186 Mfr # V31XW3
COMPARE
3 6.5 115
₹185,000.00INR / Each
Ohaus V31XW6 Valor 3000 Xtreme Portion Control Washdown Scale, 6000g x 2g; Ip-65/Nema 1101187 Mfr # V31XW6
COMPARE
6 13 115
₹185,000.00INR / Each