Ohaus Valor 2000 XW Xtreme Portion Control Washdown Scales
Enhance safety and productivity by combining rapid response and durability in one scale
  • Thick polycarbonate keypad prevents damage from impacts and punctures
  • Flow Thru™ water-resistant design is rated IPX8, channeling liquids away from internal components
  • Fast response and large dual displays allow more than one operator to use the scale at the same time
  • 304 stainless steel models add protection and checkweighing mode
  • NSF certified for food processing
  • Built-in rechargeable battery provides 50 hours continuous use
  • Spring-loaded power cord receptacle cover underneath scale protects connection from moisture
The Valor 2000 XW model features 304 stainless steel for both the housing and the platform, and are programmed to perform both weighing and check weighing functions. The check weighing mode utilizes bright red, yellow, and green LEDs to make filling, sorting, grading, and portioning easy and fast.
All models are designed with the Flow Thru™ water-resistant system, which channels liquids away from internal components and out through drain holes. This design includes silicone sealed internal components and connections to protect sensitive instrumentation from condensation.
166,000.00 - ₹194,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Ohaus V22XWE1501T Valor 2000 XW Compact Bench Scale 1, 500g x 0.2g 1161121 Mfr # V22XWE1501T
COMPARE
1.5 3 100 to 240
₹166,000.00INR / Each
Ohaus V22XWE3T Valor 2000 XW Compact Bench Scale 3, 000g x 0.5G 1161122 Mfr # V22XWE3T
COMPARE
3 6 100 to 240
₹194,000.00INR / Each
Ohaus V22XWE15T Valor 2000 XW Compact Bench Scale 15, 000g x 2g 1161124 Mfr # V22XWE15T
COMPARE
15 30 100 to 240
₹166,000.00INR / Each