Ohaus Ranger 4000 Counting Scales
Industrial construction, accurate weighing and counting
  • Stainless steel platform and die-cast aluminum housing protect internal components
  • Bright white LED display with checkweighing LED and audible indicators function in loud and dim settings
  • Fast stabilization rate increases efficiency and productivity
The Ranger 4000 bench scales feature user-friendly SmarText™ software, making scale set up fast and easy. Standard functions include weighing, parts counting, checkweighing, checkcounting, and accumulation.
Standard RS-232 interface communicates with PC or printer. Upgrade communcations with an additional RS-232, USB, or Ethernet (order separately). Increase throughput by connecting a second weighing platform (order separately). Compact printer simplifies reporting requirements (order separately).
310,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Ohaus Ranger 4000 RC41M3 Compact Counting Scale 3000 g x 0.1 g 1101555 Mfr # RC41M3
COMPARE
Compact Counting Scale 3000 g x 0.1 g
₹310,000.00INR / Each
Ohaus Ranger 4000 RC41M6  Compact Counting Scale 6000 g x 0.2 g 1101556 Mfr # RC41M6
COMPARE
Compact Counting Scale 6000 g x 0.2 g
₹310,000.00INR / Each
Ohaus Ranger 4000 RC41M30 Compact Counting Scale 30 kg x 1 g 1101558 Mfr # RC41M30
COMPARE
Compact Counting Scale 30 kg x 1 g
₹310,000.00INR / Each