Ohaus® PR Series NTEP-Approved Analytical Balances
Reliable performance at an affordable price
  • Bright easy-to-read backlit LCD
  • RS-232 interface allows connection to printer or PC
  • Small footprint saves desktop space
  • Selectable environmental filters compensate for harsh environments
  • Removable stainless steel pan and glass draft shield for easy cleaning
  • NTEP Legal-For-Trade and Measurement Canada approved
The NTEP-certified analytical balances offer basic weighing, parts counting, and percent weighing applications. A backlit LCD and easy-to-use interface make these balances simple to operate. A standard RS-232 port provides communication with point-of-sale systems or printers.
The balances are designed with a durable ABS housing and include a stainless steel pan and glass draft shield with sliding top door, which are removable for easy cleaning. Other features include an integrated weigh-below hook, security bracket, calibration lock, auto-tare, auto-dim, user-selectable span calibration points, software lockout and reset menu, user-selectable communication settings and data print options, user-definable project and user IDs, software overload/underload indicator, and stability indicator.
340,000.00 - ₹503,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Ohaus PR323N/E Analytical Balance, NTEP Approved; 320 g x 0.001 g 1180370 Mfr # PR323N/E
COMPARE
External 320 0.001
₹340,000.00INR / Each
Ohaus PR523N Analytical Balance, NTEP Approved; 520 g x 0.001 g (Internal Calibration) 1180371 Mfr # PR523N
COMPARE
Internal 520 0.001
₹503,000.00INR / Each
Ohaus PR523N/E Analytical Balance, NTEP Approved; 520 g x 0.001 g 1180372 Mfr # PR523N/E
COMPARE
External 520 0.001
₹434,000.00INR / Each