Ohaus PR Series Analytical Balances
Reliable performance at an affordable price
  • Bright, easy-to-read backlit LCD
  • RS-232 interface allows connection to printer or PC
  • Small footprint saves desktop space
  • Selectable environmental filters compensate for harsh environments
  • Removable stainless-steel pan for easy cleaning
These analytical balances offer basic weighing, parts counting, and percent weighing applications. A backlit LCD and easy-to-use interface make these balances simple to operate. A standard RS-232 port provides communication with point-of-sale systems or printers.
The balances are designed with durable ABS housing and include a stainless-steel pan and glass draft shield with sliding top door, which are removable for easy cleaning. Additional features include an integrated weigh-below hook, security bracket, calibration lock, auto-tare, auto-dim, user-selectable span calibration points, software lockout and reset menu, user-selectable communication settings and data print options, user-definable project and user IDs, software overload/underload indicator, and stability indicator.
Designed for accuracy and repeatability in weighting applications, these balances are ideal for laboratory, industrial, and educational settings at an economical price.
480,000.00 - ₹717,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Ohaus PR64/E Analytical Balance, 62g  x 0.0001g, External Calibration 1160117 Mfr # PR64/E
COMPARE
External 62 0.001
₹480,000.00INR / Each
Ohaus PR224 Analytical Balance, 220g  x 0.0001g,  Internal Calibration 1160118 Mfr # PR224
COMPARE
Internal 220 0.001
₹691,000.00INR / Each
Ohaus PR224/E Analytical Balance, 220g  x 0.0001g,  External Calibration 1160119 Mfr # PR224/E
COMPARE
External 220 0.001
₹717,000.00INR / Each
Ohaus PR124 Analytical Balance, 120g  x 0.0001g,  Internal Calibration 1160120 Mfr # PR124
COMPARE
Internal 120 0.001
₹623,000.00INR / Each
Ohaus PR124/E Analytical Balance, 120g  x 0.0001g,  External Calibration 1160121 Mfr # PR124/E
COMPARE
External 120 0.001
₹645,000.00INR / Each