Ohaus® Pioneer® PX Toploading Balances
Outstanding accuracy and repeatability at an affordable price
  • Bright backlit dual-line LCD is easy to view in any environment
  • Durable construction with metal base, ABS top housing, and stainless-steel weighing pan
  • Standard RS-232 and USB ports for easy connectivity to printers and other devices
  • Four quick keys allow for easy operation of multiple application modes.
The PX Pioneer® toploading balances deliver high weighing accuracy and repeatability at an affordable price for use in laboratory, industrial, and education applications. A bright backlit LCD provides a clear view of weighing results while a second line display offers additional information and guidance.
Standard RS-232 and USB ports make transferring results to a PC or other peripherals a simple process. The balances are equipped with a security lock to prevent theft. A standard in-use cover provides added protection against spillage and minimizes damage to the display and keypad. Other features include four user selectable operating languages (English, Chinese, Japanese, and Korean), real time clock, GMP/GLP data fields, level indicator, and menu lock. Application modes include basic weighing, parts counting, percent weighing, animal weighing, and density determination.
209,000.00 - ₹491,000.00INR / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Ohaus PX1602/E Pioneer Toploading Balance, 1600g x 0.01g, External Calibration WW-11611-87 Mfr # PX1602/E
COMPARE
External 1600 0.01
₹225,000.00INR / Each
Ohaus PX1602 Pioneer Toploading Balance, 1600g x 0.01g, Internal Calibration WW-11611-88 Mfr # PX1602
COMPARE
Internal 1600 0.01
₹279,000.00INR / Each
Ohaus PX2202/E Pioneer Toploading Balance, 2200g x 0.01g, External Calibration WW-11611-89 Mfr # PX2202/E
COMPARE
External 2200 0.01
₹247,000.00INR / Each
Ohaus PX2202 Pioneer Toploading Balance, 2200g x 0.01g, Internal Calibration WW-11611-90 Mfr # PX2202
COMPARE
Internal 2200 0.01
₹303,000.00INR / Each
Ohaus PX3202/E Pioneer Toploading Balance, 3200g x 0.01g, External Calibration WW-11611-91 Mfr # PX3202/E
COMPARE
External 3200 0.01
₹296,000.00INR / Each
Ohaus PX3202  Pioneer Toploading Balance, 3200g x 0.01g, Internal Calibration WW-11611-92 Mfr # PX3202
COMPARE
Internal 3200 0.01
₹356,000.00INR / Each
Ohaus PX4202/E Pioneer Toploading Balance, 4200g x 0.01g, External Calibration WW-11611-93 Mfr # PX4202/E
COMPARE
External 4200 0.01
₹337,000.00INR / Each
Ohaus PX4202  Pioneer Toploading Balance, 4200g x 0.01g, Internal Calibration WW-11611-94 Mfr # PX4202
COMPARE
Internal 4200 0.01
₹399,000.00INR / Each
Ohaus PX2201/E Pioneer Toploading Balance, 2200g x 0.1g, External Calibration WW-11611-95 Mfr # PX2201/E
COMPARE
External 2200 0.1
₹209,000.00INR / Each
Ohaus PX2201 Pioneer Toploading Balance, 2200g x 0.1g, Internal Calibration WW-11611-96 Mfr # PX2201
COMPARE
Internal 2200 0.1
₹258,000.00INR / Each
Ohaus PX4201/E Pioneer Toploading Balance, 4200g x 0.1g, External Calibration WW-11611-97 Mfr # PX4201/E
COMPARE
External 4200 0.1
₹241,000.00INR / Each
Ohaus PX4201  Pioneer Toploading Balance, 4200g x 0.1g, Internal Calibration WW-11611-98 Mfr # PX4201
COMPARE
Internal 4200 0.1
₹292,000.00INR / Each
Ohaus PX623 Pioneer Toploading Balance 600g x .001g, Internal Calibration WW-55000-40 Mfr # PX623
COMPARE
Internal 600 0.001
₹393,000.00INR / Each
Ohaus PX623/E Pioneer Toploading Balance 600g x .001g, External Calibration WW-55000-41 Mfr # PX623/E
COMPARE
External 600 0.001
₹344,000.00INR / Each
Ohaus PX6202 Pioneer Toploading Balance 6200g x .01g, Internal Calibration WW-55000-42 Mfr # PX6202
COMPARE
Internal 6200 0.01
₹491,000.00INR / Each
Ohaus PX6202/E Pioneer Toploading Balance 6200g x .01g, External Calibration WW-55000-43 Mfr # PX6202/E
COMPARE
External 6200 0.01
₹425,000.00INR / Each
Showing 1- 16 of 16