Ohaus Overhead Mechanical Balances
Get stable readings three times faster than other three-beam balances
  • Designed with a hanging pan system
  • Offer 10 times the readability of standard triple-beam balances
  • Feature a self-aligning beam, floating agate bearings, and counter-balancing knob for fast zeroing
  • Dial-O-Gram® balance comes equipped with a convenient, direct-reading dial calibration
  • Magnetic damping speeds up weighing times
106,000.00 - ₹129,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (g) Description
Availability Pricing
Ohaus 311-00 Cent-O-Gram Overhead Mechanical Balance, 311g x 0.01g 0100000 Mfr # 311-00
COMPARE
311 0.01 Cent-O-Gram Overhead Mechanical Balance, 311g x 0.01g
₹106,000.00INR / Each
Ohaus 310-00 Dial-O-Gram Overhead Mechanical Balance, 310g x 0.01g 0133000 Mfr # 310-00
COMPARE
310 0.01 Dial-O-Gram Overhead Mechanical Balance, 310g x 0.01g
₹129,000.00INR / Each