Ohaus Guardian 5000 Magnetic Stirrers
Ensures safe, accurate stirring of your sample
  • Precise stirring from 50 to 1800 rpm
  • Easy to read LCD display
  • Chemical resistant housing
  • IP32 ingress protection
The Guardian 5000 magnetic stirrers are designed to provide powerful mixing capabilities for use in laboratories, pharmacies, schools, businesses, and light industry. Key features combine safety, ease of use, and versatility to ensure reliable performance for everyday applications. Precise stirring from 50 to 1800 rpm is made possible by the design of a powerful motor, strong magnet, and software-controlled ramp rate.
The SmartRate™ system allows you to select stirring speeds from fast to slow, providing flexibility for various applications. To ensure samples are not over stirred, the count-down timer will automatically switch off once it reaches the set time.
A bright LCD display features speed settings, intuitive icons, and clear status indicators for easy monitoring across the lab. Other features include a RS-232 port for remote control or data logging, green indicator lights when unit is stirring, single point calibration icon, rotary knob control, two built-in support rod holders, large dip angle front panel for easy monitoring from a distance, smooth enclosure surfaces for easy cleaning, audible alarm, IP32 ingress protection, and chemical resistant housing.
135,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
High Speed (rpm) Max Stirring Volume (Liters) Power (VAC)
Availability Pricing
Ohaus Guardian 5000 e-G52ST07C  Magnetic Stirrer, 7" x 7", Ceramic; 120 VAC 8524113 Mfr # 30541635
COMPARE
1800 20 120
₹135,000.00INR / Each
Ohaus Guardian 5000 e-G52ST07C  Magnetic Stirrer, 7" x 7", Ceramic; 230 VAC 8524114 Mfr # 30541636
COMPARE
1800 20 230
₹135,000.00INR / Each