Ohaus® Explorer Plus™ EXP Series NTEP Precision Toploading Balances
Superb accuracy, seamless set up, intuitive interface, and advanced communication capabilities in one unit to deliver outstanding weighing performance with NTEP certification
  • Achieve consistent results with built-in automatic calibration
  • Manage weighing on the intuitive 7” glass touchscreen display
  • Two touchless sensors—set to activate print, zero, tare, or calibrate
  • Motorized leveling system ensures a smooth and straightforward setup
  • Detachable terminal adds flexibility
  • RS-232, USB, and Ethernet connections provide seamless data transfer
  • Supports HID connection to PC or keyboard without drivers
  • Save up to 3000 application sample libraries to easily manage multiple sample profiles
  • Power saving features include auto dim, auto standby, and sleep mode
  • Full metal housing offers durability
  • NTEP legal-for-trade and Measurement Canada approved for regulated weighing applications
Explorer Plus™ EXP precision toploading balances with NTEP Certification enhance your workflow with the outstanding weighing performance, intuitive touchscreen display, and advanced communication capabilities. Automatic, user-triggered, and span calibration options ensure precise measurements; the built-in automatic calibration performs self-calibration using an internal mass. A built-in motorized leveling system with a digital bubble automatically levels the balance with the push of a button for a smooth setup. Easily manage weighing operations on the 7” touchscreen display. An easy-to-clean, chemical-resistant glass panel displays vibrant colors for easy viewing. Detachable terminal offers flexibility—ideal for small labs or placing outside fume hoods. Set terminal status light to indicate overload or underload. Touchless sensors located on the terminal activate often used functions such as print, zero, tare, or calibrate, simply wave your hand over the sensors to activate. Built-in RS-232, USB (A and C), and Ethernet connections provide seamless data transfer; order an optional dongle for Bluetooth® and Wi-Fi capabilities. Save and recall up to 3000 sample library records to simplify your weighing operation. Each record may contain minimum weight, sample name, sample ID, batch ID, project name, and more. Order an optional fingerprint scanner for added security and a convenient and secure way to log in.
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (g)
Availability Pricing
Ohaus EXP223N/AD Explorer Plus™ EXP Series NTEP Precision Top Loading  Balance, 220g x 1mg, Auto-Draftshield and Touch Screen 2060535 Mfr # EXP223N/AD
COMPARE
220 0.001
₹1,294,000.00INR / Each
Ohaus EXP423N/AD Explorer Plus™ EXP Series NTEP Precision Top Loading  Balance, 420g x 1mg, Auto-Draftshield and Touch Screen 2060536 Mfr # EXP423N/AD
COMPARE
420 0.001
₹1,512,000.00INR / Each
Ohaus EXP623N/AD Explorer Plus™ EXP Series NTEP Precision Top Loading  Balance, 620g x 1mg, Auto-Draftshield and Touch Screen 2060537 Mfr # EXP623N/AD
COMPARE
620 0.001
₹1,848,000.00INR / Each
Ohaus EXP1203N/AD Explorer Plus™ EXP Series NTEP Precision Top Loading  Balance, 1200g x 1mg, Auto-Draftshield and Touch Screen 2060538 Mfr # EXP1203N/AD
COMPARE
1200 0.001
₹2,165,000.00INR / Each
Ohaus EXP2202N Explorer Plus™ EXP Series NTEP Precision Top Loading  Balance, 2200g x 0.01g, Touch Screen 2060539 Mfr # EXP2202N
COMPARE
2200 0.01
₹1,347,000.00INR / Each
Ohaus EXP4202N Explorer Plus™ EXP Series NTEP Precision Top Loading  Balance, 4200g x 0.01g, Touch Screen 2060540 Mfr # EXP4202N
COMPARE
4200 0.01
₹1,549,000.00INR / Each
Ohaus EXP6202N Explorer Plus™ EXP Series NTEP Precision Top Loading  Balance, 6200g x 0.01g, Touch Screen 2060541 Mfr # EXP6202N
COMPARE
6200 0.01
₹1,758,000.00INR / Each
Ohaus EXP8202N Explorer Plus™ EXP Series NTEP Precision Top Loading  Balance, 8200g x 0.01g, Touch Screen 2060542 Mfr # EXP8202N
COMPARE
8200 0.01
₹1,919,000.00INR / Each
Ohaus EXP12202N Explorer Plus™ EXP Series NTEP Precision Top Loading  Balance, 12200g x 0.01g, Touch Screen 2060543 Mfr # EXP12202N
COMPARE
12200 0.01
₹2,084,000.00INR / Each
Ohaus EXP24001N Explorer Plus™ EXP Series NTEP High-Capacity Top Loading  Balance, 24000g x 0.1g, Touch Screen, 14 Application Modes 2060544 Mfr # EXP24001N
COMPARE
24000 0.1
₹2,004,000.00INR / Each
Ohaus EXP35001N Explorer Plus™ EXP Series NTEP High-Capacity Top Loading Balance, 35000g x 0.1g, Touch Screen, 14 Application Modes 2060545 Mfr # EXP35001N
COMPARE
35000 0.1
₹2,353,000.00INR / Each
Ohaus EXP65001N Explorer Plus™ EXP Series NTEP High-Capacity Top Loading Balance, 65000g x 0.1g, Touch Screen, 14 Application Modes 2060546 Mfr # EXP65001N
COMPARE
65000 0.1
Call For Price
Showing 1- 12 of 12