Ohaus® Explorer™ EXR Series Analytical Balances
High-performance balances offer intuitive interface and outstanding accuracy
  • Achieve consistent results with built-in automatic calibration
  • Delivers 0.1 mg repeatability and 0.06 mg linearity for outstanding accuracy and reliability
  • Anti-static draft shield minimizes static electricity around the balance
  • Manage weighing on the intuitive 7” glass touchscreen display
  • Two touchless sensors—set to activate print, zero, tare, or calibrate
  • Digital leveling bubble ensures a smooth and straightforward setup
  • RS-232, USB, and Ethernet connections provide seamless data transfer
  • Supports HID connection to PC or keyboard without drivers
  • Save up to 1000 application sample libraries to easily manage multiple sample profiles
  • Power saving features include auto dim, auto standby, and sleep mode
  • Full metal housing offers durability
Enhance your workflow with the Explorer™ EXR analytical balances featuring outstanding weighing performance, intuitive touchscreen display, and advanced communication capabilities. Automatic, user-triggered, and span calibration options ensure precise measurements; the built-in automatic calibration performs self-calibration using an internal mass. Select from 11 application modes: Basic Weighing, Parts Counting, Check Counting, Percent Weighing, Check Weighing, Dynamic Weighing, Totalization, Formulation, Differential, Density Determination, Peak Hold.
Easily manage weighing operations on the 7” touchscreen display. An easy-to-clean, chemical-resistant glass panel displays vibrant colors for easy viewing. Removable draft shield offers hassle-free cleaning and long-term care. Anti-static coating on the draft shield reduces static electricity around the balance ensuring accurate results. Touchless sensors located on the terminal activate often used functions such as print, zero, tare, or calibrate, simply wave your hand over the sensors to activate.
Built-in RS-232, USB (A and C), and Ethernet connections provide seamless data transfer; order an optional dongle for Bluetooth® and Wi-Fi capabilities. Save and recall up to 1000 sample library records to simplify your weighing operation. Each record may contain minimum weight, sample name, sample ID, batch ID, project name, and more.
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g)
Availability Pricing
Ohaus EXR124 Explorer™ EXR Series Analytical Balance, 120g x 0.1mg, Draftshield, Touch Screen 2060517 Mfr # EXR124
COMPARE
120
₹1,688,000.00INR / Each
Ohaus EXR224 Explorer™ EXR Series Analytical Balance, 220g x 0.1mg, Draftshield, Touch Screen 2060518 Mfr # EXR224
COMPARE
220
₹1,873,000.00INR / Each
Ohaus EXR324 Explorer™ EXR Series Analytical Balance, 320g x 0.1mg, Draftshield, Touch Screen 2060519 Mfr # EXR324
COMPARE
320
₹2,200,000.00INR / Each