Ohaus Defender® 6000 Washdown Indicators
Stand up to harsh environments with IP68 immersion/ IP69K washdown protection
  • Large, bright display offers clear result for fast processing
  • Three-color bar for quick check
  • Choose between polycarbonate or 316 stainless steel housing
Ohaus Defender® 6000 washdown indicators are designed for food, chemical, and pharmaceutical applications including production, processing, and packaging. Easily read results on the 45 mm high digits.
198,000.00 - ₹272,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
USB Interface Capacity (lbs) Description
Availability Pricing
Ohaus i-DT61PW Defender Advanced Indicator, Polycarbonate Plastic 1180405 Mfr # 30575583
COMPARE
No 99999 Defender Advanced Indicator, Polycarbonate Plastic
₹198,000.00INR / Each
Ohaus i-DT61PW  Defender Advanced Indicator, Polycarbonate Plastic, Front Mount 1180406 Mfr # 30575585
COMPARE
No 99999 Defender Advanced Indicator, Polycarbonate Plastic, Front Mount
₹198,000.00INR / Each
Ohaus i-DT61XWE Defender Advanced Indicator, 316 Stainless Steel 1180407 Mfr # 30575584
COMPARE
Yes 999999 Defender Advanced Indicator, 316 Stainless Steel
₹272,000.00INR / Each