Ohaus Defender® 6000 Washdown Base
Advanced IP68/IP69K washdown base designed for food, chemical, and pharmaceutical applications
  • Stands up to harsh cleaning in processing and packing environments
  • EasyConnect™ cable allows instant configuration with indicators
  • Meets metrological approvals, is NSF certified, and supports HACCP systems
  • 150% overload capacity protection
The Defender 6000 Series washdown base handles the most demanding applications with 316 stainless steel performance and frame, and hermetically sealed stainless steel IP69K load cell. It stands up to harsh cleaning in processing and packaging environments with IP68 immersion/ IP69K washdown protection. Base is equipped with an EasyConnect cable to allow instant configuration with Defender 5000 and 6000 indicators, speeding up installation. The 316 stainless steel construction provides excellent corrosion protection in your food, chemical processing, and packaging applications. The open design minimizes dirt buildup and supports easy cleaning.
322,000.00 - ₹508,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (lbs) Readability (g) Description
Availability Pricing
Ohaus i-D5K1S Defender 6000 Washdown Base, 10 lb x 0.001 lb 1180400 Mfr # 30575603
COMPARE
10 0.5 Defender 6000 Washdown Base, 10 lb x 0.001 lb
₹322,000.00INR / Each
Ohaus i-D12K1R Defender 6000 Washdown Base, 25 lb x 0.002 lb 1180401 Mfr # 30575604
COMPARE
25 1 Defender 6000 Washdown Base, 25 lb x 0.002 lb
₹344,000.00INR / Each
Ohaus i-D25K1R Defender 6000 Washdown Base, 50 lb x 0.005 lb 1180402 Mfr # 30575605
COMPARE
50 2 Defender 6000 Washdown Base, 50 lb x 0.005 lb
₹344,000.00INR / Each
Ohaus i-D50K1L Defender 6000 Washdown Base, 100 lb x 0.01 lb 1180403 Mfr # 30575606
COMPARE
100 5 Defender 6000 Washdown Base, 100 lb x 0.01 lb
₹508,000.00INR / Each
Ohaus i-D150K1L Defender 6000 Washdown Base, 300 lb x 0.02 lb 1180404 Mfr # 30575607
COMPARE
300 10 Defender 6000 Washdown Base, 300 lb x 0.02 lb
₹436,000.00INR / Each