Ohaus Defender® 6000 Hybrid Washdown Bench Scale with Stainless Steel Indicator
Precise, versatile, and remarkably durable balance to handle your most demanding weighing applications
  • Durable stainless steel platform and frame construction, IP67 loadcell, and IP68/69K indicator
  • 316 stainless steel indicator – enhanced protection against corrosion
  • Backlit display allows for fast processing
  • Integrated leveling bubbles and adjustable nonslip rubber feet provide enhanced accuracy and stability
The Defender 6000 hybrid washdown bench scale is ideal for a multitude of applications, including industrial and commercial production and packaging. The combination of the 5000 W series washdown base and 6000 series indicator handles demanding applications. Stand up to harsh environments with IP68 immersion/IP69K protected indicator, 304 stainless steel construction, and IP67 stainless steel load cell. Large digits on the bright display quickly and clearly shows results for fast processing. The Defender 5000 base features moisture-protected single-point load cells, adjustable rubber feet, and leveling bubbles to prevent slippage and damage on wet or uneven surfaces. Bases are equipped with an EasyConnect™ cable to instantly connect to the indicator.
316 stainless steel indicator offers enhanced protection against corrosion from cleaners and additives and features a configurable bar that can be used for detailed checking of package weights to ensure compliance. It offers multiple connectivity options, including built-in RS-232 and USB host, RS-232/RS-485/USB device, analog output, Wi-Fi/Bluetooth®, Ethernet, and discrete I/O with relays. It also features menu password protection.
506,000.00 - ₹875,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Capacity (lbs) Platform Length (in) Platform Width (in)
Availability Pricing
Ohaus i-D61XWE2WQS6 Defender® 6000 Hybrid Washdown Scale, 5 lb x 0.0002 lb, 10 x 10" Base, Stainless Steel Indicator 1180423 Mfr # 30631706
COMPARE
5 10 10
₹506,000.00INR / Each
Ohaus i-D61XWE5WQS6 Defender® 6000 Hybrid Washdown Scale, 10 lb x 0.0005 lb, 10 x 10" Base, Stainless Steel Indicator 1180424 Mfr # 30631707
COMPARE
10 10 10
₹506,000.00INR / Each
Ohaus i-D61XWE12WQS6 Defender® 6000 Hybrid Washdown Scale, 25 lb x 0.001 lb, 10 x 10"Base, Stainless Steel Indicator 1180425 Mfr # 30631718
COMPARE
25 10 10
₹506,000.00INR / Each
Ohaus i-D61XWE12WQR6 Defender® 6000 Hybrid Washdown Scale, 25 lb x 0.001 lb, 12 x 12"Base, Stainless Steel Indicator 1180426 Mfr # 30631719
COMPARE
25 12 12
₹637,000.00INR / Each
Ohaus i-D61XWE25WQR6 Defender® 6000 Hybrid Washdown Scale, 50 lb x 0.002 lb, 12 x 12"Base, Stainless Steel Indicator 1180427 Mfr # 30631720
COMPARE
50 12 12
₹546,000.00INR / Each
Ohaus i-D61XWE25WQL7 Defender® 6000 Hybrid Washdown Scale, 50 lb x 0.002 lb, 18 x 18"Base, Stainless Steel Indicator 1180428 Mfr # 30631722
COMPARE
50 18 18
₹750,000.00INR / Each
Ohaus i-D61XWE50WQR6 Defender® 6000 Hybrid Washdown Scale, 100 lb x 0.005 lb, 12 x 12"Base, Stainless Steel Indicator 1180429 Mfr # 30631721
COMPARE
100 12 12
₹637,000.00INR / Each
Ohaus i-D61XWE50WQL7 Defender® 6000 Hybrid Washdown Scale, 100 lb x 0.005 lb, 18 x 18"Base, Stainless Steel Indicator 1180430 Mfr # 30631723
COMPARE
100 18 18
₹875,000.00INR / Each
Ohaus i-D61XWE125WQL7 Defender® 6000 Hybrid Washdown Scale, 250 lb x 0.01 lb, 18 x 18"Base, Stainless Steel Indicator 1180431 Mfr # 30631724
COMPARE
250 18 18
₹750,000.00INR / Each
Showing 1- 9 of 9