Ohaus Defender® 6000 Hybrid Washdown Bench Scale with Front Mounted Polycarbonate Indicator
Precise, versatile, and remarkably durable balance to handle your most demanding weighing applications
  • Durable stainless steel platform and frame construction, IP67 loadcell, and IP68/69K indicator
  • Front mounted polycarbonate indicator provides superior protection against moisture
  • Backlit display allows for fast processing
  • Integrated leveling bubbles and adjustable nonslip rubber feet provide enhanced accuracy and stability
The Defender 6000 hybrid washdown bench scale is ideal for a multitude of applications, including industrial and commercial production and packaging. The combination of the 5000 W series washdown base and 6000 series indicator handles demanding applications. Stand up to harsh environments with IP68 immersion/IP69K protected indicator, 304 stainless steel construction, and IP67 stainless steel load cell. Large digits on the bright display quickly and clearly shows results for fast processing. The Defender 5000 base features moisture-protected single-point load cells, adjustable rubber feet, and leveling bubbles to prevent slippage and damage on wet or uneven surfaces. Bases are equipped with an EasyConnect™ cable to instantly connect to the indicator.
The polycarbonate indicator housing for superior protection against moisture and features a color bar used for quick checking of package weights for fast go/no-go alerts and low battery indicator. It can store up to 1000 weighing results which can be transferred to a PC or system for traceability.
485,000.00 - ₹653,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Capacity (lbs) Platform Length (in) Platform Width (in)
Availability Pricing
Ohaus i-D61PW2WQS5 Defender® 6000 Hybrid Washdown Scale, 5 lb x 0.0002 lb, 10 x 10" Base, Front Mount Polycarbonate Indicator 1180441 Mfr # 30631725
COMPARE
5 10 10
₹485,000.00INR / Each
Ohaus i-D61PW5WQS5 Defender® 6000 Hybrid Washdown Scale, 10 lb x 0.0005 lb, 10 x 10" Base, Front Mount Polycarbonate Indicator 1180442 Mfr # 30631726
COMPARE
10 10 10
₹485,000.00INR / Each
Ohaus i-D61PW12WQS5 Defender® 6000 Hybrid Washdown Scale, 25 lb x 0.001 lb, 10 x 10" Base, Front Mount Polycarbonate Indicator 1180443 Mfr # 30631727
COMPARE
25 10 10
₹485,000.00INR / Each
Ohaus i-D61PW12WQR5 Defender® 6000 Hybrid Washdown Scale, 25 lb x 0.001 lb, 12 x 12" Base, Front Mount Polycarbonate Indicator 1180444 Mfr # 30631728
COMPARE
25 12 12
₹522,000.00INR / Each
Ohaus i-D61PW25WQR5 Defender® 6000 Hybrid Washdown Scale, 50 lb x 0.002 lb, 12 x 12" Base, Front Mount Polycarbonate Indicator 1180445 Mfr # 30631729
COMPARE
50 12 12
₹522,000.00INR / Each
Ohaus i-D61PW25WQL5 Defender® 6000 Hybrid Washdown Scale, 50 lb x 0.002 lb, 18 x 18" Base, Front Mount Polycarbonate Indicator 1180446 Mfr # 30572969
COMPARE
50 18 18
₹653,000.00INR / Each
Ohaus i-D61PW50WQR5 Defender® 6000 Hybrid Washdown Scale, 100 lb x 0.005 lb, 12 x 12" Base, Front Mount Polycarbonate Indicator 1180447 Mfr # 30631730
COMPARE
100 12 12
₹522,000.00INR / Each
Ohaus i-D61PW50WQL5 Defender® 6000 Hybrid Washdown Scale, 100 lb x 0.005 lb, 18 x 18" Base, Front Mount Polycarbonate Indicator 1180448 Mfr # 30572970
COMPARE
100 18 18
₹653,000.00INR / Each
Ohaus i-D61PW125WQL5 Defender® 6000 Hybrid Washdown Scale, 250 lb x 0.01 lb, 18 x 18" Base, Front Mount Polycarbonate Indicator 1180449 Mfr # 30572971
COMPARE
250 18 18
₹653,000.00INR / Each
Showing 1- 9 of 9