Ohaus Defender 6000 Advanced Washdown Scale with Stainless Steel Indicator
Withstand harsh use and cleaning with IP68 immersion/IP69K washdown protection
  • NSF certified, USDA-ASM accepted, and supports HACCP systems
  • Withstand harsh use and cleaning with IP68 immersion/IP69K washdown protection
  • Durable 316 stainless steel construction with hermetically-sealed load cell
  • GMP/GLP with time/date output allows for traceability of results
  • Multiple software modes to meet your food, chemical processing, and packaging needs
The Defender 6000 features a 316 stainless steel platform and frame, and an advanced washdown indicator with an IP69K stainless steel load cell, so it can endure high-pressure washdowns and extreme temperatures, water pressure, and cleaning products common in the food and chemical industries. The base has an open frame design with no exposed threads, tight corners, or other places where material can build up. The column has been designed with an open back for easy cleaning.
316 stainless steel indicator offers enhanced protection against corrosion from cleaners and additives. It also features multiple connectivity options, including built-in RS-232 and USB host, second RS-232/RS-485/USB device, analog output, Wi-Fi/Bluetooth®, Ethernet, and discrete I/O with relays. The hands-free, touchless sensor reduces contamination risk.
The Defender 6000 scale meets the weighing needs of practically any food or chemical processing application by featuring configurable checkweighing, percent weighing, and filling and dynamic weighing or display hold. The easy-to-use application modes eliminate the need for long and complicated manual calculations. Dedicated target key allows for quick setup of package over/under weights, speeding up line changeovers.
541,000.00 - ₹666,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (lbs) Readability (g) Description
Availability Pricing
Ohaus i-D61XWE5K1S6 Defender 6000 Advanced Washdown Scale, 10 lb x 0.001 lb, Stainless Steel Indicator 1180408 Mfr # 30575563
COMPARE
10 0.5 Defender 6000 Advanced Washdown Scale, 10 lb x 0.001 lb, Stainless Steel Indicator
₹541,000.00INR / Each
Ohaus i-D61XWE12K1R6 Defender 6000 Advanced Washdown Scale, 25 lb x 0.002 lb, Stainless Steel Indicator 1180409 Mfr # 30575564
COMPARE
25 1 Defender 6000 Advanced Washdown Scale, 25 lb x 0.002 lb, Stainless Steel Indicator
₹566,000.00INR / Each
Ohaus i-D61XWE25K1R6 Defender 6000 Advanced Washdown Scale, 50 lb x 0.005 lb, Stainless Steel Indicator 1180410 Mfr # 30575565
COMPARE
50 2 Defender 6000 Advanced Washdown Scale, 50 lb x 0.005 lb, Stainless Steel Indicator
₹566,000.00INR / Each
Ohaus i-D61XWE50K1L7 Defender 6000 Advanced Washdown Scale, 100 lb x 0.01 lb, Stainless Steel Indicator 1180411 Mfr # 30575566
COMPARE
100 5 Defender 6000 Advanced Washdown Scale, 100 lb x 0.01 lb, Stainless Steel Indicator
₹666,000.00INR / Each
Ohaus i-D61XWE150K1L7 Defender 6000 Advanced Washdown Scale, 300 lb x 0.02 lb, Stainless Steel Indicator 1180412 Mfr # 30575567
COMPARE
300 10 Defender 6000 Advanced Washdown Scale, 300 lb x 0.02 lb, Stainless Steel Indicator
₹666,000.00INR / Each