Ohaus® Defender™ 5000 WQ and WT Series Scale Bases
Enhanced durability with moisture-protected single-point load cells
  • Removable 304 stainless steel weighing pan offers quick cleaning
  • Designed with integrated leveling bubbles and adjustable non-slip rubber feet for enhanced accuracy and stability
These heavy-duty industrial scale bases feature a 304 stainless steel platform, making them ideal for a wide range of industrial applications. Bases feature moisture-protected single-point load cells, adjustable rubber feet, and leveling bubbles to prevent slippage and damage on wet or uneven surfaces. Bases are equipped with an EasyConnect cable to instantly connect to a Defender 5000 TD52 indicator with easy. Indicators sold separately.
Washdown bases feature all stainless steel construction with IP67 rated load cell for washdown environments.
268,000.00 - ₹776,000.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Platform Length (in) Platform Width (in)
Availability Pricing
Ohaus D2WQS Defender 5000 Stainless Steel Scale Base Only, Base Style Square , 5x0.0002lb 1180284 Mfr # D2WQS
COMPARE
2.5 10 10
₹268,000.00INR / Each
Ohaus D5WQS Defender 5000 Stainless Steel Scale Base Only, Base Style Square,  10x0.0005lb 1180285 Mfr # D5WQS
COMPARE
5 10 10
₹268,000.00INR / Each
Ohaus D12WQR Defender 5000 Stainless Steel Scale Base Only, Base Style Square, 25 lbs x 0.005lb 1180287 Mfr # D12WQR
COMPARE
12.5 12 12
₹285,000.00INR / Each
Ohaus D50WQR Defender 5000 Stainless Steel Scale Base Only, Base Style Square,  100x0.005lb 1180289 Mfr # D50WQR
COMPARE
50 12 12
₹285,000.00INR / Each
Ohaus D50WQL Defender 5000 Stainless Steel Scale Base Only, Base Style Square,  100x0.005lb 1180291 Mfr # D50WQL
COMPARE
50 18 18
₹452,000.00INR / Each
Ohaus D125WQL Defender 5000 Stainless Steel Scale Base Only, Base Style Square,  250x0.01lb 1180292 Mfr # D125WQL
COMPARE
125 18 18
₹527,000.00INR / Each
Ohaus D125WTX Defender 5000 Stainless Steel Scale Base Only, Base Style Rectangle,  250x0.01lb 1180294 Mfr # D125WTX
COMPARE
125 24 18
₹614,000.00INR / Each
Ohaus D50WQV Defender 5000 Stainless Steel Scale Base Only, Base Style Square,  100x0.005lb 1180296 Mfr # D50WQV
COMPARE
50 24 24
₹626,000.00INR / Each
Ohaus D125WQV Defender 5000 Stainless Steel Scale Base Only, Base Style Square,  250x0.01lb 1180297 Mfr # D125WQV
COMPARE
125 24 24
₹730,000.00INR / Each
Ohaus D250WQV Defender 5000 Stainless Steel Scale Base Only, Base Style Square,  500x0.02lb 1180298 Mfr # D250WQV
COMPARE
250 24 24
₹666,000.00INR / Each
Ohaus D500WQV Defender 5000 Stainless Steel Scale Base Only, Base Style Square,  1000x0.05lb 1180299 Mfr # D500WQV
COMPARE
500 24 24
₹776,000.00INR / Each
Showing 1- 11 of 11