Ohaus® Defender™ 5000 Washdown Low Profile Industrial Scales
Rugged stainless steel base construction for demanding industrial operations
  • Easily read measurements with large 5.3” LCD with white LED backlight
  • Stainless steel platform with attached indicator provide maximum durability, rigidity, and structural integrity in a small footprint
  • Features include counting, percent weighing, dynamic/animal weighing, display hold, and checkweighing modes
  • Store up to 50,000 items and 1,000 user profiles on included Micro SD card
  • Durable washdown stainless steel indicator features an IP68/NEMA 4X protection rating
  • Enhanced accuracy with ±0.01% repeatability and linearity
These heavy-duty industrial bench scales combine a large, user-friendly indicator with rugged stainless steel base for demanding industrial environments. Ideal for shipping, receiving, production, packaging, and general commercial weighing applications. The indicator attached to the front of the base allows for the compact scale to fit easily on benchtops.
The next generation Defender 5000 series bench scales feature a maximum counting resolution to 1:500,000, an easy-to-operate alpha-numeric keypad and function keys, and RS-232 output.
517,000.00 - ₹1,101,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Platform Length (in) Platform Width (in)
Availability Pricing
Ohaus D52XW50WQR5 Defender 5000 Low Profile Washdown Scale, Base Style Square ,  100x0.005lb 1180316 Mfr # D52XW50WQR5
COMPARE
50 12 12
₹559,000.00INR / Each
Ohaus D52XW25WQL5 Defender 5000 Low Profile Washdown Scale, Base Style Square ,  50x0.002lb 1180317 Mfr # D52XW25WQL5
COMPARE
25 18 18
₹823,000.00INR / Each
Ohaus D52XW50WQL5 Defender 5000 Low Profile Washdown Scale, Base Style Square ,  100x0.005lb 1180318 Mfr # D52XW50WQL5
COMPARE
50 18 18
₹706,000.00INR / Each
Ohaus D52XW125WQL5 Defender 5000 Low Profile Washdown Scale, Base Style Square ,  250x0.01lb 1180319 Mfr # D52XW125WQL5
COMPARE
125 18 18
₹823,000.00INR / Each
Ohaus D52XW2WQS5 Defender 5000 Low Profile Washdown Scale, Base Style Square ,  5x0.0002lb 1180320 Mfr # D52XW2WQS5
COMPARE
2.5 10 10
₹517,000.00INR / Each
Ohaus D52XW12WQS5 Defender 5000 Low Profile Washdown Scale, Base Style Square ,  25x0.001lb 1180322 Mfr # D52XW12WQS5
COMPARE
12.5 10 10
₹517,000.00INR / Each
Ohaus D52XW12WQR5 Defender 5000 Low Profile Washdown Scale, Base Style Square ,  25x0.001lb 1180323 Mfr # D52XW12WQR5
COMPARE
12.5 12 12
₹559,000.00INR / Each
Ohaus D52XW25WQR5 Defender 5000 Low Profile Washdown Scale, Base Style Square ,  50x0.002lb 1180324 Mfr # D52XW25WQR5
COMPARE
25 12 12
₹652,000.00INR / Each
Ohaus D52XW50WQV5 Defender 5000 Low Profile Washdown Scale, Base Style Square ,  100x0.005lb 1180328 Mfr # D52XW50WQV5
COMPARE
50 24 24
₹1,101,000.00INR / Each
Showing 1- 9 of 9