Ohaus® Defender™ 5000 Washdown Industrial Scales
Rugged stainless steel construction for demanding harsh environment industrial operations
  • Easily read measurements with large 5.3” LCD with white LED backlight
  • Stainless steel platform, steel frames, and industrial load cells provide maximum durability, rigidity, and structural integrity
  • Features include counting, percent weighing, dynamic/animal weighing, display hold, and checkweighing modes
  • Store up to 50,000 items and 1,000 user profiles on included Micro SD card
  • Enhanced accuracy with ±0.01% repeatability and linearity
These heavy-duty industrial bench scales combine a large, user-friendly indicator with rugged stainless steel base for demanding industrial environments. Ideal for shipping, receiving, production, packaging, and general commercial weighing applications.
The next generation Defender 5000 series bench scales feature a maximum counting resolution to 1:500,000, an easy-to-operate alpha-numeric keypad and function keys, and RS-232 output.
The washdown indicator is constructed of 304 stainless steel with sand blasting surface treatment, making this ideal for harsh environment industrial applications. The indicator features an IP68 and NEMA 4X protection rating.
545,000.00 - ₹1,189,000.00INR / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Platform Length (in) Platform Width (in)
Availability Pricing
Ohaus D52XW2WQS6 Defender 5000 Washdown Scale, Base Style Square , 5x0.0002lb 1180300 Mfr # D52XW2WQS6
COMPARE
2.5 10 10
₹588,000.00INR / Each
Ohaus D52XW5WQS6 Defender 5000 Washdown Scale, Base Style Square , 10x0.0005lb 1180301 Mfr # D52XW5WQS6
COMPARE
5 10 10
₹565,000.00INR / Each
Ohaus D52XW12WQR6 Defender 5000 Washdown Scale, Base Style Square , 25x0.001lb 1180303 Mfr # D52XW12WQR6
COMPARE
12.5 12 12
₹636,000.00INR / Each
Ohaus D52XW25WQR6 Defender 5000 Washdown Scale, Base Style Square , 50x0.002lb 1180304 Mfr # D52XW25WQR6
COMPARE
25 12 12
₹545,000.00INR / Each
Ohaus D52XW50WQR6 Defender 5000 Washdown Scale, Base Style Square , 100x0.005lb 1180305 Mfr # D52XW50WQR6
COMPARE
50 12 12
₹636,000.00INR / Each
Ohaus D52XW25WQL7 Defender 5000 Washdown Scale, Base Style Square , 50x0.002lb 1180306 Mfr # D52XW25WQL7
COMPARE
25 18 18
₹769,000.00INR / Each
Ohaus D52XW50WQL7 Defender 5000 Washdown Scale, Base Style Square , 100x0.005lb 1180307 Mfr # D52XW50WQL7
COMPARE
50 18 18
₹897,000.00INR / Each
Ohaus D52XW125WQL7 Defender 5000 Washdown Scale, Base Style Square , 250x0.01lb 1180308 Mfr # D52XW125WQL7
COMPARE
125 18 18
₹769,000.00INR / Each
Ohaus D52XW50WTX7 Defender 5000 Washdown Scale, Base Style Rectangle , 100x0.005lb 1180309 Mfr # D52XW50WTX7
COMPARE
50 24 18
₹847,000.00INR / Each
Ohaus D52XW50WQV8 Defender 5000 Washdown Scale, Base Style Square , 100x0.005lb 1180312 Mfr # D52XW50WQV8
COMPARE
50 24 24
₹1,145,000.00INR / Each
Ohaus D52XW125WQV8 Defender 5000 Washdown Scale, Base Style Square , 250x0.01lb 1180313 Mfr # D52XW125WQV8
COMPARE
125 24 24
₹1,159,000.00INR / Each
Ohaus D52XW250WQV8 Defender 5000 Washdown Scale, Base Style Square , 500x0.02lb 1180314 Mfr # D52XW250WQV8
COMPARE
250 24 24
₹1,189,000.00INR / Each
Ohaus D52XW500WQV8 Defender 5000 Washdown Scale, Base Style Square , 1000x0.05lb 1180315 Mfr # D52XW500WQV8
COMPARE
500 24 24
₹1,189,000.00INR / Each
Showing 1- 13 of 13