Ohaus® Defender™ 5000 RQ and RT Series Scale Bases
Enhanced durability with moisture-protected single-point load cells
  • Removable 304 stainless steel weighing pan offers quick cleaning
  • Designed with integrated leveling bubbles and adjustable non-slip rubber feet for enhanced accuracy and stability
These heavy-duty industrial scale bases feature a 304 stainless steel platform, making them ideal for a wide range of industrial applications. Bases feature moisture-protected single-point load cells, adjustable rubber feet, and leveling bubbles to prevent slippage and damage on wet or uneven surfaces. Bases are equipped with an EasyConnect cable to instantly connect to a Defender 5000 TD52 indicator with easy. Indicators sold separately.
Standard bases feature sand grey powder coated steel frame and aluminum load cell.
235,000.00 - ₹463,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Platform Length (in) Platform Width (in)
Availability Pricing
Ohaus D25RQR Defender 5000 Scale Base Only, Base Style Square,  50x0.002lb 1180267 Mfr # D25RQR
COMPARE
25 12 12
₹235,000.00INR / Each
Ohaus D50RQR Defender 5000 Scale Base Only, Base Style Square,  100x0.005lb 1180268 Mfr # D50RQR
COMPARE
50 12 12
₹235,000.00INR / Each
Ohaus D12RTR Defender 5000 Scale Base Only, Base Style Rectangle,  25x0.001lb 1180269 Mfr # D12RTR
COMPARE
12.5 14 12
₹268,000.00INR / Each
Ohaus D25RTR Defender 5000 Scale Base Only, Base Style Rectangle,  50x0.002lb 1180270 Mfr # D25RTR
COMPARE
25 14 12
₹268,000.00INR / Each
Ohaus D50RTR Defender 5000 Scale Base Only, Base Style Rectangle,  100x0.005lb 1180271 Mfr # D50RTR
COMPARE
50 14 12
₹268,000.00INR / Each
Ohaus D50RQL Defender 5000 Scale Base Only, Base Style Square,  100x0.005lb 1180272 Mfr # D50RQL
COMPARE
50 18 18
₹336,000.00INR / Each
Ohaus D125RQL Defender 5000 Scale Base Only, Base Style Square,  250x0.01lb 1180273 Mfr # D125RQL
COMPARE
125 18 18
₹393,000.00INR / Each
Ohaus D125RTX Defender 5000 Scale Base Only, Base Style Rectangle,  250x0.01lb 1180275 Mfr # D125RTX
COMPARE
125 24 18
₹352,000.00INR / Each
Ohaus D50RQV Defender 5000 Scale Base Only, Base Style Square,  100x0.005lb 1180277 Mfr # D50RQV
COMPARE
50 24 24
₹397,000.00INR / Each
Ohaus D250RQV Defender 5000 Scale Base Only, Base Style Square,  500x0.02lb 1180279 Mfr # D250RQV
COMPARE
250 24 24
₹463,000.00INR / Each
Showing 1- 10 of 10