Ohaus® Defender™ 5000 Low Profile Industrial Scales with Stainless Steel Indicator
Rugged stainless steel base construction for demanding industrial operations
  • Easily read measurements with large 5.3” LCD with white LED backlight
  • Stainless steel platform with attached indicator provide maximum durability, rigidity, and structural integrity in a small footprint
  • Features include counting, percent weighing, dynamic/animal weighing, display hold, and checkweighing modes
  • Store up to 50,000 items and 1,000 user profiles on included Micro SD card
  • Durable stainless steel indicator features an IP68/NEMA 4X protection rating
  • Enhanced accuracy with ±0.01% repeatability and linearity
These heavy-duty industrial bench scales combine a large, user-friendly indicator with rugged stainless steel base for demanding industrial environments. Ideal for shipping, receiving, production, packaging, and general commercial weighing applications. The indicator attached to the front of the base allows for the compact scale to fit easily on benchtops.
The next generation Defender 5000 series bench scales feature a maximum counting resolution to 1:500,000, an easy-to-operate alpha-numeric keypad and function keys, and RS-232 output.
490,000.00 - ₹652,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Platform Length (in) Platform Width (in)
Availability Pricing
Ohaus D52XW12RQR5 Defender 5000 Low Profile Scale with S/S Indicator, Base Style Square , 25x0.001lb 1180215 Mfr # D52XW12RQR5
COMPARE
12.5 12 12
₹490,000.00INR / Each
Ohaus D52XW50RQR5 Defender 5000 Low Profile Scale with S/S Indicator, Base Style Square , 100x0.005lb 1180217 Mfr # D52XW50RQR5
COMPARE
50 12 12
₹490,000.00INR / Each
Ohaus D52XW25RTR5 Defender 5000 Low Profile Scale with S/S Indicator, Base Style Rectangle , 50x0.002lb 1180219 Mfr # D52XW25RTR5
COMPARE
25 14 12
₹593,000.00INR / Each
Ohaus D52XW50RTR5 Defender 5000 Low Profile Scale with S/S Indicator, Base Style Rectangle , 100x0.005lb 1180220 Mfr # D52XW50RTR5
COMPARE
50 14 12
₹508,000.00INR / Each
Ohaus D52XW50RQL5 Defender 5000 Low Profile Scale with S/S Indicator, Base Style Square , 100x0.005lb 1180221 Mfr # D52XW50RQL5
COMPARE
50 18 18
₹585,000.00INR / Each
Ohaus D52XW125RQL5 Defender 5000 Low Profile Scale with S/S Indicator, Base Style Square , 250x0.01lb 1180222 Mfr # D52XW125RQL5
COMPARE
125 18 18
₹585,000.00INR / Each
Ohaus D52XW250RTX5 Defender 5000 Low Profile Scale with S/S Indicator, Base Style Rectangle , 500x0.02lb 1180225 Mfr # D52XW250RTX5
COMPARE
250 24 18
₹620,000.00INR / Each
Ohaus D52XW125RQV5 Defender 5000 Low Profile Scale with S/S Indicator, Base Style Square , 250x0.01lb 1180227 Mfr # D52XW125RQV5
COMPARE
125 24 24
₹652,000.00INR / Each
Ohaus D52XW250RQV5 Defender 5000 Low Profile Scale with S/S Indicator, Base Style Square , 500x0.02lb 1180229 Mfr # D52XW250RQV5
COMPARE
250 24 24
₹652,000.00INR / Each
Showing 1- 9 of 9