Ohaus® Defender™ 5000 Low Profile Industrial Scales with ABS Plastic Indicator
Rugged stainless steel base construction for demanding industrial operations
  • Easily read measurements with large 5.3” LCD with white LED backlight
  • Stainless steel platform with attached indicator provide maximum durability, rigidity, and structural integrity in a small footprint
  • Features include counting, percent weighing, dynamic/animal weighing, display hold, and checkweighing modes
  • Store up to 50,000 items and 1,000 user profiles on included Micro SD card
  • Indicator is constructed of ABS plastic — ideal for economical applications
  • Enhanced accuracy with ±0.01% repeatability and linearity
These heavy-duty industrial bench scales combine a large, user-friendly indicator with rugged stainless steel base for demanding industrial environments. Ideal for shipping, receiving, production, packaging, and general commercial weighing applications. The indicator attached to the front of the base allows for the compact scale to fit easily on benchtops.
The next generation Defender 5000 series bench scales feature a maximum counting resolution to 1:500,000, an easy-to-operate alpha-numeric keypad and function keys, and RS-232 output.
426,000.00 - ₹584,000.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Platform Length (in) Platform Width (in)
Availability Pricing
Ohaus D52P12RQR5 Defender 5000 Low Profile Scale, Base Style Square , 25x0.001lb 1180200 Mfr # D52P12RQR5
COMPARE
12.5 12 12
₹426,000.00INR / Each
Ohaus D52P25RQR5 Defender 5000 Low Profile Scale, Base Style Square , 50x0.002lb 1180201 Mfr # D52P25RQR5
COMPARE
25 12 12
₹426,000.00INR / Each
Ohaus D52P50RQR5 Defender 5000 Low Profile Scale, Base Style Square , 100x0.005lb 1180202 Mfr # D52P50RQR5
COMPARE
50 12 12
₹426,000.00INR / Each
Ohaus D52P12RTR5 Defender 5000 Low Profile Scale, Base Style Rectangle , 25x0.001lb 1180203 Mfr # D52P12RTR5
COMPARE
12.5 14 12
₹444,000.00INR / Each
Ohaus D52P25RTR5 Defender 5000 Low Profile Scale, Base Style Rectangle , 50x0.002lb 1180204 Mfr # D52P25RTR5
COMPARE
25 14 12
₹444,000.00INR / Each
Ohaus D52P50RTR5 Defender 5000 Low Profile Scale, Base Style Rectangle , 100x0.005lb 1180205 Mfr # D52P50RTR5
COMPARE
50 14 12
₹444,000.00INR / Each
Ohaus D52P50RQL5 Defender 5000 Low Profile Scale, Base Style Square , 100x0.005lb 1180206 Mfr # D52P50RQL5
COMPARE
50 18 18
₹522,000.00INR / Each
Ohaus D52P125RQL5 Defender 5000 Low Profile Scale, Base Style Square , 250x0.01lb 1180207 Mfr # D52P125RQL5
COMPARE
125 18 18
₹522,000.00INR / Each
Ohaus D52P125RTX5 Defender 5000 Low Profile Scale, Base Style Rectangle , 250x0.01lb 1180209 Mfr # D52P125RTX5
COMPARE
125 24 18
₹546,000.00INR / Each
Ohaus D52P50RQV5 Defender 5000 Low Profile Scale, Base Style Square, 100x0.005lb 1180211 Mfr # D52P50RQV5
COMPARE
50 24 24
₹584,000.00INR / Each
Ohaus D52P250RQV5 Defender 5000 Low Profile Scale, Base Style Square , 500x0.02lb 1180213 Mfr # D52P250RQV5
COMPARE
250 24 24
₹584,000.00INR / Each
Showing 1- 11 of 11