Ohaus® Defender™ 5000 Industrial Scales with Stainless Steel Indicator
Rugged stainless steel construction for demanding industrial operations
  • Easily read measurements with large 5.3” LCD with white LED backlight
  • Stainless steel platform, steel frames, and industrial load cells provide maximum durability, rigidity, and structural integrity
  • Features include counting, percent weighing, dynamic/animal weighing, display hold, and checkweighing modes
  • Store up to 50,000 items and 1,000 user profiles on included Micro SD card
  • Enhanced accuracy with ±0.01% repeatability and linearity
These heavy-duty industrial bench scales combine a large, user-friendly indicator with rugged stainless steel base for demanding industrial environments. Ideal for shipping, receiving, production, packaging, and general commercial weighing applications.
The next generation Defender 5000 series bench scales feature a maximum counting resolution to 1:500,000, an easy-to-operate alpha-numeric keypad and function keys, and RS-232 output.
Indicator is constructed of 304 stainless steel with sand blasting surface treatment, making this ideal for rugged industrial applications. The indicator features an IP68 and NEMA 4X protection rating.
517,000.00 - ₹877,000.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Platform Length (in) Platform Width (in)
Availability Pricing
Ohaus D52XW12RQR1 Defender 5000 Scale with S/S Indicator, Base Style Square , 25x0.001lb 1180248 Mfr # D52XW12RQR1
COMPARE
12.5 12 12
₹517,000.00INR / Each
Ohaus D52XW25RQR1 Defender 5000 Scale with S/S Indicator, Base Style Square , 50x0.002lb 1180249 Mfr # D52XW25RQR1
COMPARE
25 12 12
₹517,000.00INR / Each
Ohaus D52XW50RQR1 Defender 5000 Scale with S/S Indicator, Base Style Square , 100x0.005lb 1180250 Mfr # D52XW50RQR1
COMPARE
50 12 12
₹517,000.00INR / Each
Ohaus D52XW12RTR1 Defender 5000 Scale with S/S Indicator, Base Style Rectangle , 25x0.001lb 1180251 Mfr # D52XW12RTR1
COMPARE
12.5 12 14
₹517,000.00INR / Each
Ohaus D52XW25RTR1 Defender 5000 Scale with S/S Indicator, Base Style Rectangle , 50x0.002lb 1180252 Mfr # D52XW25RTR1
COMPARE
25 12 14
₹517,000.00INR / Each
Ohaus D52XW50RTR1 Defender 5000 Scale with S/S Indicator, Base Style Rectangle , 100x0.005lb 1180253 Mfr # D52XW50RTR1
COMPARE
50 12 14
₹517,000.00INR / Each
Ohaus D52XW50RQL2 Defender 5000 Scale with S/S Indicator, Base Style Square , 100x0.005lb 1180254 Mfr # D52XW50RQL2
COMPARE
50 18 18
₹722,000.00INR / Each
Ohaus D52XW250RTX2 Defender 5000 Scale with S/S Indicator, Base Style Rectangle , 500x0.02lb 1180258 Mfr # D52XW250RTX2
COMPARE
250 18 24
₹653,000.00INR / Each
Ohaus D52XW50RQV3 Defender 5000 Scale with S/S Indicator, Base Style Square , 100x0.005lb 1180259 Mfr # D52XW50RQV3
COMPARE
50 24 24
₹706,000.00INR / Each
Ohaus D52XW125RTV3 Defender 5000 Scale with S/S Indicator, Base Style Rectangle , 250x0.01lb 1180263 Mfr # D52XW125RTV3
COMPARE
125 23 19/32 31 1/2
₹877,000.00INR / Each
Ohaus D52XW500RTV3 Defender 5000 Scale with S/S Indicator, Base Style Rectangle , 1000x0.05lb 1180265 Mfr # D52XW500RTV3
COMPARE
500 23 19/32 31 1/2
₹877,000.00INR / Each
Showing 1- 11 of 11