Ohaus® Defender™ 5000 Industrial Scales with ABS Plastic Indicator
Rugged stainless steel base construction for demanding industrial operations
  • Easily read measurements with large 5.3” LCD with white LED backlight
  • Stainless steel platform, steel frames, and industrial load cells provide maximum durability, rigidity, and structural integrity
  • Features include counting, percent weighing, dynamic/animal weighing, display hold, and checkweighing modes
  • Store up to 50,000 items and 1,000 user profiles on included Micro SD card
  • Enhanced accuracy with ±0.01% repeatability and linearity
These heavy-duty industrial bench scales combine a large, user-friendly indicator with rugged stainless steel base for demanding industrial environments. Ideal for shipping, receiving, production, packaging, and general commercial weighing applications.
The next generation Defender 5000 series bench scales feature a maximum counting resolution to 1:500,000, an easy-to-operate alpha-numeric keypad and function keys, and RS-232 output.
Indicator is durable and economical, constructed of ABS plastic, making it ideal for rugged applications at a practical price.
463,000.00 - ₹685,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Platform Length (in) Platform Width (in)
Availability Pricing
Ohaus D52P12RQR1 Defender 5000 Scale, Base Style Square, 25x0.001lb 1180230 Mfr # D52P12RQR1
COMPARE
12.5 12 12
₹540,000.00INR / Each
Ohaus D52P25RQR1 Defender 5000 Scale, Base Style Square, 50x0.002lb 1180231 Mfr # D52P25RQR1
COMPARE
25 12 12
₹463,000.00INR / Each
Ohaus D52P50RQR1 Defender 5000 Scale, Base Style Square, 100x0.005lb 1180232 Mfr # D52P50RQR1
COMPARE
50 12 12
₹463,000.00INR / Each
Ohaus D52P25RTR1 Defender 5000 Scale, Base Style Rectangle, 50x0.002lb 1180234 Mfr # D52P25RTR1
COMPARE
25 12 14
₹463,000.00INR / Each
Ohaus D52P50RTR1 Defender 5000 Scale, Base Style Rectangle, 100x0.005lb 1180235 Mfr # D52P50RTR1
COMPARE
50 12 14
₹463,000.00INR / Each
Ohaus D52P50RQL2 Defender 5000 Scale, Base Style Square, 100x0.005lb 1180236 Mfr # D52P50RQL2
COMPARE
50 18 18
₹565,000.00INR / Each
Ohaus D52P125RQL2 Defender 5000 Scale, Base Style Square, 250x0.01lb 1180237 Mfr # D52P125RQL2
COMPARE
125 18 18
₹565,000.00INR / Each
Ohaus D52P50RTX2 Defender 5000 Scale, Base Style Rectangle, 100x0.005lb 1180238 Mfr # D52P50RTX2
COMPARE
50 18 24
₹596,000.00INR / Each
Ohaus D52P125RTX2 Defender 5000 Scale, Base Style Rectangle, 250x0.01lb 1180239 Mfr # D52P125RTX2
COMPARE
125 18 24
₹596,000.00INR / Each
Ohaus D52P125RQV3 Defender 5000 Scale, Base Style Square, 250x0.01lb 1180242 Mfr # D52P125RQV3
COMPARE
125 24 24
₹685,000.00INR / Each
Showing 1- 10 of 10