Ohaus® Defender® 3000 Washdown Bench Scale with Column-Mounted SS Indicator
Rugged design for industrial weighing, counting, totalization, and checkweighing applications in wet environments
  • IP67-rated load cell allows hose down for easy cleaning
  • View measurements on large, backlit LCD screen
  • Column-mount IP-66-rated indicators are easy to view even from a distance
  • 304 SS indicators, platform, frame, column, and mounting bracket offer excellent durability
  • Green-yellow-red color change indicator in checkweighing mode
  • Built-in bidirectional RS-232 for printing or data connectivity
  • Enhanced accuracy with ±1d repeatability and linearity
The Defender 3000 washdown bench scales provide durable, reliable weighing to make your job easier. Scales feature 304 stainless steel (SS) platform, frame, and IP66-rated indicator and IP67-rated load cell to endure damp work environments while allowing a complete washdown if needed. Indicator comes with a 304 SS bracket and internal rechargeable battery as a standard.
Easily view the readings on a large 6-digit, 7-segment LCD with four selectable background colors—amber, green, red, or yellow. Large 1.8" (45-mm) high digits are easily seen even from a distance. Bidirectional RS-232 interface allow connection to a printer or computer for easy data transfer; optional second port USB or Ethernet connection is available.
Use the scales for routine weighing, counting, totalization and checkweighing applications. Convenient checkweighing mode ensures the weight is within your target range. Quality control is easy with intuitive color change—green, yellow, or red—indicates whether the weight meets the target. Checkweighing mode supports regular positive checkweighing into a package, negative checkweighing out of a bin or tote, and zero checkweighing + or - against a sample weight to suit your weighing needs.
446,000.00 - ₹621,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Height (in) Platform Length (in)
Availability Pricing
Ohaus D33XW15C1R6 Defender® 3000 Washdown Scale, Column-Mount SS Indicator, 14"x12" Platform, 15kg x 2g 1134839 Mfr # D33XW15C1R6
COMPARE
15 21 5/16 14
₹446,000.00INR / Each
Ohaus D33XW30C1R6 Defender® 3000 Washdown Scale, Column-Mount SS Indicator, 14"x12" Platform, 30kg x 5g 1134840 Mfr # D33XW30C1R6
COMPARE
30 21 5/16 14
₹446,000.00INR / Each
Ohaus D33XW75C1L7 Defender® 3000 Washdown Scale, Column-Mount SS Indicator, 21.7"x16.5" Platform, 75kg x 10g 1134841 Mfr # D33XW75C1L7
COMPARE
75 35 5/16 21 3/4
₹504,000.00INR / Each
Ohaus D33XW75C1R6 Defender® 3000 Washdown Scale, Column-Mount SS Indicator, 14"x12" Platform, 75kg x 10g 1134842 Mfr # D33XW75C1R6
COMPARE
75 21 5/16 14
₹446,000.00INR / Each
Ohaus D33XW150C1L7 Defender® 3000 Washdown Scale, Column-Mount SS Indicator, 21.7"x16.5" Platform, 150kg x 20g 1134843 Mfr # D33XW150C1L7
COMPARE
150 35 5/16 21 3/4
₹504,000.00INR / Each
Ohaus D33XW150C1X7 Defender® 3000 Washdown Scale, Column-Mount SS Indicator, 25.6"x19.7" Platform, 150kg x 20g 1134844 Mfr # D33XW150C1X7
COMPARE
150 35 1/4 25 5/8
₹621,000.00INR / Each
Ohaus D33XW300C1X7 Defender® 3000 Washdown Scale, Column-Mount SS Indicator, 25.6"x19.7" Platform, 300kg x 50g 1134845 Mfr # D33XW300C1X7
COMPARE
300 35 1/4 25 5/8
₹621,000.00INR / Each
Showing 1- 7 of 7