Ohaus® Defender® 3000 Indicators
Precise, versatile, and remarkably durable balance indicators to weighing applications
  • Large 6-digit, 7-segment LCD with multi-color backlight provides easy-to-read measurements
  • Designed with bi-directional RS-232 interface for printing or data connectivity
  • NTEP Class III/IIIL certified
Ohaus® Defender® 3000 indicators are designed with multiple application modes including Weighing, Counting, Checkweighing, and Totalization. Easily read results on the large multicolor backlit LCD with 45 mm high digits.
Indicators are available with a durable ABS housing and powder coated steel mounting bracket or with an IP66 stainless steel housing and mounting bracket.
102,000.00 - ₹131,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Battery Material Description
Availability Pricing
Ohaus i-DT33P Defender® 3000 Indicator, ABS Plastic; 100-240 VAC, 50/60 Hz 1134821 Mfr # DT33P
COMPARE
Optional: C Cell or rechargeable battery 11348-94 sold separately ABS Defender® 3000 Indicator, ABS Plastic; 100-240 VAC, 50/60 Hz
₹102,000.00INR / Each
Ohaus i-DT33XW Defender® 3000 Indicator, Washdown, Stainless Steel; 100-240 VAC, 50/60 Hz 1134822 Mfr # DT33XW
COMPARE
1 Rechargeable Seal Lead-Acid (included) 304 Stainless Steel Defender® 3000 Indicator, Washdown, Stainless Steel; 100-240 VAC, 50/60 Hz
₹131,000.00INR / Each