Ohaus® Defender® 3000 Hybrid Bench Scale with Front-Mounted SS Indicator
Rugged design for industrial weighing, counting, totalization, and checkweighing applications
  • View measurements on large, backlit LCD screen
  • Front-mount IP66-rated indicators are easy to view even from a distance
  • Green-yellow-red color change indicator in checkweighing mode
  • Built-in bidirectional RS-232 for printing or data connectivity
  • Enhanced accuracy with ±1d repeatability and linearity
The Defender 3000 hybrid bench scales provide durable, reliable weighing to make your job easier. Scales feature 304 stainless steel (SS) platform, bracket, handle, and IP66-rated indicator; powder-coated base; and aluminum load cell, making them ideal for production, warehouse, and shipping and receiving areas. Indicator comes with a 304 SS carrying handle and internal rechargeable battery as a standard.
Easily view the readings on a large 6-digit, 7-segment LCD with four selectable background colors—amber, green, red, or yellow. Large 1.8" (45-mm) high digits are easily seen even from a distance. Bidirectional RS-232 interface allow connection to a printer or computer for easy data transfer; optional second port USB or Ethernet connection is available.
Use the scales for routine weighing, counting, totalization and checkweighing applications. Convenient checkweighing mode ensures the weight is within your target range. Quality control is easy with intuitive color change—green, yellow, or red—indicates whether the weight meets the target. Checkweighing mode supports regular positive checkweighing into a package, negative checkweighing out of a bin or tote, and zero checkweighing + or - against a sample weight to suit your weighing needs.
261,000.00 - ₹336,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Height (in) Platform Length (in)
Availability Pricing
Ohaus D33XW15B1R5 Defender® 3000 Hybrid Bench Scale, Front-Mount SS Indicator, 14"x12" Platform, 15kg x 2g 1134862 Mfr # D33XW15B1R5
COMPARE
15 4 13/16 12
₹261,000.00INR / Each
Ohaus D33XW30B1R5 Defender® 3000 Hybrid Bench Scale, Front-Mount SS Indicator, 14"x12" Platform, 30kg x 5g 1134863 Mfr # D33XW30B1R5
COMPARE
30 4 13/16 12
₹261,000.00INR / Each
Ohaus D33XW75B1L5 Defender® 3000 Hybrid Bench Scale, Front-Mount SS Indicator, 21.7"x16.5" Platform, 75kg x 10g 1134864 Mfr # D33XW75B1L5
COMPARE
75 5 5/16 16 1/2
₹311,000.00INR / Each
Ohaus D33XW75B1R5 Defender® 3000 Hybrid Bench Scale, Front-Mount SS Indicator, 14"x12" Platform, 75kg x 10g 1134865 Mfr # D33XW75B1R5
COMPARE
75 4 13/16 12
₹261,000.00INR / Each
Ohaus D33XW150B1L5 Defender® 3000 Hybrid Bench Scale, Front-Mount SS Indicator, 21.7"x16.5" Platform, 150kg x 20g 1134866 Mfr # D33XW150B1L5
COMPARE
150 5 5/16 16 1/2
₹311,000.00INR / Each
Ohaus D33XW150B1X5 Defender® 3000 Hybrid Bench Scale, Front-Mount SS Indicator, 25.6"x19.7" Platform, 150kg x 20g 1134867 Mfr # D33XW150B1X5
COMPARE
150 5 7/8 19 3/4
₹336,000.00INR / Each
Ohaus D33XW300B1X5 Defender® 3000 Hybrid Bench Scale, Front-Mount SS Indicator, 25.6"x19.7" Platform, 300kg x 50g 1134868 Mfr # D33XW300B1X5
COMPARE
300 5 7/8 19 3/4
₹336,000.00INR / Each
Showing 1- 7 of 7