Ohaus® Defender® 3000 Hybrid Bench Scale with Column-Mounted SS Indicator
Rugged design for industrial weighing, counting, totalization, and checkweighing applications
  • View measurements on large, backlit LCD screen
  • Column-mount IP-66-rated indicators are easy to view even from a distance
  • Green-yellow-red color change indicator in checkweighing mode
  • Built-in bidirectional RS-232 for printing or data connectivity
  • Enhanced accuracy with ±1d repeatability and linearity
The Defender 3000 hybrid bench scales provide durable, reliable weighing to make your job easier. Scales feature 304 stainless steel (SS) platform, bracket, and IP66-rated indicator; powder-coated steel base and column; and aluminum load cell, making them ideal for production, warehouse, and shipping and receiving areas. Indicator comes with a 304 SS bracket and internal rechargeable battery as a standard.
Easily view the readings on a large 6-digit, 7-segment LCD with four selectable background colors—amber, green, red, or yellow. Large 1.8" (45-mm) high digits are easily seen even from a distance. Bidirectional RS-232 interface allow connection to a printer or computer for easy data transfer; optional second port USB or Ethernet connection is available.
Use the scales for routine weighing, counting, totalization and checkweighing applications. Convenient checkweighing mode ensures the weight is within your target range. Quality control is easy with intuitive color change—green, yellow, or red—indicates whether the weight meets the target. Checkweighing mode supports regular positive checkweighing into a package, negative checkweighing out of a bin or tote, and zero checkweighing + or - against a sample weight to suit your weighing needs.
274,000.00 - ₹372,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Height (in) Platform Length (in)
Availability Pricing
Ohaus D33XW15B1R1 Defender® 3000 Hybrid Bench Scale, Column-Mount SS Indicator, 14"x12" Platform, 15kg x 2g 1134853 Mfr # D33XW15B1R1
COMPARE
15 21 5/16 12
₹274,000.00INR / Each
Ohaus D33XW30B1R1 Defender® 3000 Hybrid Bench Scale, Column-Mount SS Indicator, 14"x12" Platform, 30kg x 5g 1134854 Mfr # D33XW30B1R1
COMPARE
30 21 5/16 12
₹274,000.00INR / Each
Ohaus D33XW75B1L2 Defender® 3000 Hybrid Bench Scale, Column-Mount SS Indicator, 21.7"x16.5" Platform, 68cm Column, 75kg x 10g 1134855 Mfr # D33XW75B1L2
COMPARE
75 35 5/16 16 1/2
₹322,000.00INR / Each
Ohaus D33XW75B1R1 Defender® 3000 Hybrid Bench Scale, Column-Mount SS Indicator, 21.7"x16.5" Platform, 33cm Column, 75kg x 10g 1134856 Mfr # D33XW75B1R1
COMPARE
75 35 5/16 12
₹274,000.00INR / Each
Ohaus D33XW150B1L2 Defender® 3000 Hybrid Bench Scale, Column-Mount SS Indicator, 21.7"x16.5" Platform, 150kg x 20g 1134857 Mfr # D33XW150B1L2
COMPARE
150 35 5/16 16 1/2
₹322,000.00INR / Each
Ohaus D33XW150B1X2 Defender® 3000 Hybrid Bench Scale, Column-Mount SS Indicator, 25.6"x19.7" Platform, 150kg x 20g 1134858 Mfr # D33XW150B1X2
COMPARE
150 35 5/16 19 3/4
₹348,000.00INR / Each
Ohaus D33XW300B1X2 Defender® 3000 Hybrid Bench Scale, Column-Mount SS Indicator, 25.6"x19.7" Platform, 300kg x 50g 1134859 Mfr # D33XW300B1X2
COMPARE
300 35 5/16 19 3/4
₹348,000.00INR / Each
Ohaus D33XW300B1V3 Defender® 3000 Hybrid Bench Scale, Column-Mount SS Indicator, 31.5"x23.6" Platform, 300kg x 50g 1134860 Mfr # D33XW300B1V3
COMPARE
300 47 23 5/8
₹372,000.00INR / Each
Ohaus D33XW600B1V3 Defender® 3000 Hybrid Bench Scale, Column-Mount SS Indicator, 31.5"x23.6" Platform, 600kg x 100g 1134861 Mfr # D33XW600B1V3
COMPARE
600 47 23 5/8
₹372,000.00INR / Each
Showing 1- 9 of 9