Ohaus® Defender® 3000 Bench Scale with Front Mounted ABS Indicator
Precise, versatile, and remarkably durable balance to handle your weighing applications
  • Large 6-digit, 7-segment LCD with multi-color backlight provides easy-to-read measurements
  • Designed with bi-directional RS-232 interface for printing or data connectivity
  • Durable 304 stainless steel platform and front-mount bracket with handle, powder-coated steel base, and aluminum load cell
  • Front mounted indicator features a durable ABS housing
  • Enhanced accuracy with ±1d repeatability and linearity
The Defender ® 3000 bench scale features a simple, yet rugged, front mounted indicator design with tactile keys, backlit LCD, and bi-directional RS-232 interface. Designed with multiple application modes including Weighing, Counting, Checkweighing, and Totalization makes these scales ideal for general weighing, simple counting and checkweighing applications in production, packaging, warehouse, inventory, shipping and receiving areas.
209,000.00 - ₹311,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Platform Width (in) Readability (g)
Availability Pricing
Ohaus i-D33P15B1R5 Defender® 3000 Scale, Front Mount ABS Indicator, 14" x 12" Base, 15 kg x 2 g; 100-240 VAC, 50/60 Hz 1134832 Mfr # D33P15B1R5
COMPARE
15 12 2
₹244,000.00INR / Each
Ohaus i-D33P30B1R5 Defender® 3000 Scale, Front Mount ABS Indicator, 14" x 12" Base, 30 kg x 5 g; 100-240 VAC, 50/60 Hz 1134833 Mfr # D33P30B1R5
COMPARE
30 12 5
₹244,000.00INR / Each
Ohaus i-D33P75B1L5 Defender® 3000 Scale, Front Mount ABS Indicator, 21.7" x 16.5" Base, 75 kg x 10 g; 100-240 VAC, 50/60 Hz 1134834 Mfr # D33P75B1L5
COMPARE
75 16 1/2 10
₹242,000.00INR / Each
Ohaus i-D33P75B1R5 Defender® 3000 Scale, Front Mount ABS Indicator, 14" x 12" Base, 75 kg x 10 g; 100-240 VAC, 50/60 Hz 1134835 Mfr # D33P75B1R5
COMPARE
75 12 10
₹209,000.00INR / Each
Ohaus i-D33P150B1L5 Defender® 3000 Scale, Front Mount ABS Indicator, 21.7" x 16.5" Base, 150 kg x 20 g; 100-240 VAC, 50/60 Hz 1134836 Mfr # D33P150B1L5
COMPARE
150 16 1/2 20
₹281,000.00INR / Each
Ohaus i-D33P150B1X5 Defender® 3000 Scale, Front Mount ABS Indicator, 25.6" x 19.7" Base, 150 kg x 20 g; 100-240 VAC, 50/60 Hz 1134837 Mfr # D33P150B1X5
COMPARE
150 19 45/64 20
₹311,000.00INR / Each
Ohaus i-D33P300B1X5 Defender® 3000 Scale, Front Mount ABS Indicator, 25.6" x 19.7" Base, 300 kg x 50 g; 100-240 VAC, 50/60 Hz 1134838 Mfr # D33P300B1X5
COMPARE
300 19 45/64 50
₹311,000.00INR / Each
Showing 1- 7 of 7