Ohaus® Defender® 3000 Bench Scale with Column Mounted ABS Indicator
Precise, versatile, and remarkably durable balance to handle your weighing applications
  • Large 6-digit, 7-segment LCD with multi-color backlight provides easy-to-read measurements
  • Designed with bi-directional RS-232 interface for printing or data connectivity
  • Durable 304 stainless steel platform, powder-coated steel tubular frame and column, and aluminum load cell
  • Indicator features a durable ABS housing and powder coated steel mounting bracket
  • Enhanced accuracy with ±1d repeatability and linearity
The Defender ® 3000 bench scale features a simple, yet rugged, tubular-frame base design and indicators with tactile keys, backlit LCD, and bi-directional RS-232 interface. Designed with multiple application modes including Weighing, Counting, Checkweighing, and Totalization makes these scales ideal for general weighing, simple counting and checkweighing applications in production, packaging, warehouse, inventory, shipping and receiving areas.
246,000.00 - ₹348,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Display Column Height (in) Readability (g)
Availability Pricing
Ohaus i-D33P15B1R1 Defender® 3000 Scale, 13" Column Mount ABS Indicator, 14" x 12" Base, 15 kg x 2 g; 100-240 VAC, 50/60 Hz 1134823 Mfr # D33P15B1R1
COMPARE
15 13 2
₹246,000.00INR / Each
Ohaus i-D33P30B1R1 Defender® 3000 Scale, 13" Column Mount ABS Indicator, 14" x 12" Base, 30 kg x 5 g; 100-240 VAC, 50/60 Hz 1134824 Mfr # D33P30B1R1
COMPARE
30 13 5
₹246,000.00INR / Each
Ohaus i-D33P75B1L2 Defender® 3000 Scale, 26.8" Column Mount ABS Indicator, 21.7" x 16.5" Base, 75 kg x 10 g; 100-240 VAC, 50/60 Hz 1134825 Mfr # D33P75B1L2
COMPARE
75 26 51/64 10
₹297,000.00INR / Each
Ohaus i-D33P75B1R1 Defender® 3000 Scale, 13" Column Mount ABS Indicator, 14" x 12" Base, 75 kg x 10 g; 100-240 VAC, 50/60 Hz 1134826 Mfr # D33P75B1R1
COMPARE
75 13 10
₹246,000.00INR / Each
Ohaus i-D33P150B1L2 Defender® 3000 Scale, 26.8" Column Mount ABS Indicator, 21.7" x 16.5" Base, 150 kg x 20 g; 100-240 VAC, 50/60 Hz 1134827 Mfr # D33P150B1L2
COMPARE
150 26 51/64 20
₹297,000.00INR / Each
Ohaus i-D33P150B1X2 Defender® 3000 Scale, 26.8" Column Mount ABS Indicator, 25.6" x 19.7" Base, 150 kg x 20 g; 100-240 VAC, 50/60 Hz 1134828 Mfr # D33P150B1X2
COMPARE
150 26 51/64 20
₹322,000.00INR / Each
Ohaus i-D33P300B1X2 Defender® 3000 Scale, 26.8" Column Mount ABS Indicator, 25.6" x 19.7" Base, 300 kg x 50 g; 100-240 VAC, 50/60 Hz 1134829 Mfr # D33P300B1X2
COMPARE
300 26 51/64 50
₹322,000.00INR / Each
Ohaus i-D33P300B1V3 Defender® 3000 Scale, 38.6" Column Mount ABS Indicator, 31.5" x 23.6" Base, 300 kg x 50 g; 100-240 VAC, 50/60 Hz 1134830 Mfr # D33P300B1V3
COMPARE
300 38 19/32 50
₹348,000.00INR / Each
Ohaus i-D33P600B1V3 Defender® 3000 Scale, 38.6" Column Mount ABS Indicator, 31.5" x 23.6" Base, 600 kg x 100 g; 100-240 VAC, 50/60 Hz 1134831 Mfr # D33P600B1V3
COMPARE
600 38 19/32 100
₹348,000.00INR / Each
Showing 1- 9 of 9