Ohaus Adventurer AX NTEP Approved Toploading Balances
Advanced features and a wide weighing capacity range provide incredible versatility
  • GLP/GMP compliance via real-time clock and recording of sample, project name, and balance ID
  • Two USB ports (one on the front and one on the back) provide connectivity with both a PC and a portable external device at the same time
  • Cloning feature saves settings to USB flash drive for transfer to other Adventurer balances
  • Color touch screen offers easy and fast operation with or without gloves
  • Six mechanical keys that provide tactile feedback allow the operator to perform repetitive tasks
  • lluminated level indicator located on the front panel makes adjustment easy in any lighting conditions
  • Choose between internal and external calibration models
  • Advanced weighing functions include: part counting, percent, dynamic, density, check weighing, totalization, and formulation modes
Additional features include RS-232 interface, integral weigh below hook, full housing in-use cover, removable stainless steel pan, die-cast metal bottom housing, security bracket, illuminated up-front level indicator, four adjustable feet, software lockout menus, stability indicator, software overload/underload indicators, user selectable environmental settings, audible indicator, user selectable brightness settings, auto dim, touch screen calibration, auto tare, user selectable operating language.
298,000.00 - ₹565,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Ohaus AX223N/E Adventurer Toploading Balance 220g x0.001g NTEP WW-11019-08 Mfr # AX223N/E
COMPARE
220 100 to 240 0.0001
₹386,000.00INR / Each
Ohaus AX423N Adventurer Toploading Balance 420g x 0.001g NTEP WW-11019-11 Mfr # AX423N
COMPARE
420 100 to 240 0.0001
₹565,000.00INR / Each
Ohaus AX423N/E Adventurer Toploading Balance 420g x 0.001g NTEP WW-11019-12 Mfr # AX423N/E
COMPARE
420 100 to 240 0.0001
₹460,000.00INR / Each
Ohaus AX523N/E Adventurer Toploading Balance 520g x 0.001g NTEP WW-11019-15 Mfr # AX523N/E
COMPARE
520 100 to 240 0.0001
₹511,000.00INR / Each
Ohaus AX622N/E Adventurer Toploading Balance 620g x 0.01g NTEP WW-11019-18 Mfr # AX622N/E
COMPARE
620 100 to 240 0.01
₹298,000.00INR / Each
Ohaus AX1502N/E Adventurer Toploading Balance 1, 500g x 0.01g NTEP WW-11019-21 Mfr # AX1502N/E
COMPARE
1500 100 to 240 0.01
₹337,000.00INR / Each
Ohaus AX4202N/E Adventurer Toploading Balance 4, 200g x 0.01g NTEP WW-11019-27 Mfr # AX4202N/E
COMPARE
4200 100 to 240 0.01
₹474,000.00INR / Each
Ohaus AX8201N/E Adventurer Toploading Balance 8, 200g x 0.1g NTEP WW-11019-33 Mfr # AX8201N/E
COMPARE
8200 100 to 240 0.1
₹374,000.00INR / Each
Showing 1- 8 of 8