North Silver Shield Gloves
Use alone or as a chemically resistant liner within a rubberized glove
These flexible lightweight gloves are made of a five-layer polyethylene and ethylene vinyl alcohol copolymerconstruction that resists permeation by a wide range of aromatic, chlorinated, aliphatic solvents, ketones, and esters. Ideal for lab or industrial use. Ambidextrous.
37,300.00INR / Pkg of 10
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Size
Availability Pricing
Honeywell SSG/11-12 North Gloves, large, pack of 10 pair 8635802 Mfr # SSG/11
COMPARE
Large
₹37,300.00INR / Pkg of 10