Honeywell SSG/11-12 North Gloves, large, pack of 10 pair
Representative image only.
Honeywell SSG/11-12 North Gloves, large, pack of 10 pair
Honeywell | Mfr: SSG/11| Item: 8635802
Use alone or as a chemically resistant liner within a rubberized glove
₹37,300.00INR / Pkg of 10
₹37,300.00INR / Pkg of 10
Product Description
More About this Item
These flexible lightweight gloves are made of a five-layer polyethylene and ethylene vinyl alcohol copolymerconstruction that resists permeation by a wide range of aromatic, chlorinated, aliphatic solvents, ketones, and esters. Ideal for lab or industrial use. Ambidextrous.
Conformite Europeenne
Specifications
  • MaterialNorfoil®
  • SizeLarge
  • ColorGray
  • SterileNo
  • DescriptionGloves, large, pack of 10 pair