Nordson Divert™ 3-Way Stopcocks with Luer Fittings, Polycarbonate
Easily control the flow of a liquid or gas using an on/off diverter
  • Low-torque handle provides smooth adjustments
  • Diverter arrows indicate off position
  • DEHP free
  • ISO 80369-7 compliant
Add a stopcock to your system for an easy on/off control of your fluid. Each three-way stopcock features two female Luer fittings plus a male Luer fitting with or without a lock. Luer fittings on stopcocks ensure a quick, leak-free connection. Diverter arrow indicates off position, enhancing operational efficiency and minimizing errors. Transparent polycarbonate body makes it easy to see the flow.
600.00 - ₹10,100.00INR / Pkg of 25
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Luer Connection Type (Port 3) Max Operating Pressure (PSI) Quantity
Availability Pricing
Nordson Divert 3-Way Stopcock, Polycarbonate, Female Luer x Female Luer x Male Luer; 1/Ea 5614084 Mfr # FCVFFM-008
COMPARE
Male Luer 50 1
₹680.00INR / Each
Nordson Divert 3-Way Stopcock, Polycarbonate, Female Luer x Female Luer x Locking Male Luer; 1/Ea 5614087 Mfr # FCVFFMR-008
COMPARE
Locking Male Luer 50 1
₹600.00INR / Each
Nordson Divert 3-Way Stopcock, High Pressure, Polycarbonate, Female Luer x Female Luer x Locking Male Luer; 1/Ea 5614090 Mfr # FCVHPFFMR-008
COMPARE
Locking Male Luer 500 1
₹980.00INR / Each
Nordson Divert 3-Way Stopcock, Polycarbonate, Female Luer x Female Luer x Male Luer; 25/Pk 5614092 Mfr # FCVFFM-008
COMPARE
Male Luer 50 25
₹10,100.00INR / Pkg of 25
Nordson Divert 3-Way Stopcock, Polycarbonate, Female Luer x Female Luer x Locking Male Luer; 25/Pk 5614095 Mfr # FCVFFMR-008
COMPARE
Locking Male Luer 50 25
₹8,700.00INR / Pkg of 25
Nordson Divert 3-Way Stopcock, High Pressure, Polycarbonate, Female Luer x Female Luer x Locking Male Luer; 25/Pk 5614098 Mfr # FCVHPFFMR-008
COMPARE
Locking Male Luer 500 25
₹7,700.00INR / Pkg of 25