Nordson Divert 3-Way Stopcock, Polycarbonate, Female Luer x Female Luer x Locking Male Luer; 1/Ea
Representative image only.
Nordson Divert 3-Way Stopcock, Polycarbonate, Female Luer x Female Luer x Locking Male Luer; 1/Ea
| Mfr: FCVFFMR-008| Item: 5614087
Easily control the flow of a liquid or gas using an on/off diverter
₹600.00INR / Each
₹600.00INR / Each
Product Description
Key Features
  • Low-torque handle provides smooth adjustments
  • Diverter arrows indicate off position
  • DEHP free
  • ISO 80369-7 compliant
  • HPDE diverter
  • Female Luer x Female Luer x Locking Male Luer
  • 1 each
  • Manufacturer defect warranty
More About this Item
Add a stopcock to your system for an easy on/off control of your fluid. Each three-way stopcock features two female Luer fittings plus a male Luer fitting with or without a lock. Luer fittings on stopcocks ensure a quick, leak-free connection. Diverter arrow indicates off position, enhancing operational efficiency and minimizing errors. Transparent polycarbonate body makes it easy to see the flow.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPolycarbonate
  • Number Of Ports3
  • Connection TypeLuer x Luer x Locking Luer
  • Max Operating Pressure (PSI)50
  • Regulatory ApprovalsNon-DEHP
  • Luer Connection Type (Port 1)Female Luer
  • Luer Connection Type (Port 2)Female Luer
  • Luer Connection Type (Port 3)Locking Male Luer
  • Quantity1
  • Description3-Way Stopcock, Polycarbonate, Female Luer x Female Luer x Locking Male Luer; 1/Ea
  • WarrantyManufacturer defect