Nordson Divert™ 3-Way Stopcocks with Bond Port, Polycarbonate
Easily control the flow of a liquid or gas using an on/off diverter
  • Low-torque handle provides smooth adjustments
  • Diverter arrow indicates off position
  • DEHP free
  • ISO 80369-7 compliant
Add a stopcock to your system for an easy on/off control of your fluid. Three-way stopcocks feature two female Luer fittings plus a bond port. Luer fittings on stopcocks ensure a quick, leak-free connection; bond port creates a permanent connection, simply match the tube OD to bond port size. Diverter arrow indicates off position, enhancing operational efficiency and minimizing errors. Transparent polycarbonate body makes it easy to see the flow.
600.00 - ₹9,700.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Bond Port Quantity
Availability Pricing
Nordson Divert 3-Way Stopcock, Polycarbonate, Female Luer x Female Luer x 0.110" Bond Port; 1/Ea 5614085 Mfr # FCVFFM-030
COMPARE
0.110" 1
₹600.00INR / Each
Nordson Divert 3-Way Stopcock, Polycarbonate, Female Luer x Female Luer x 0.130" Bond Port; 1/Ea 5614086 Mfr # FCVFFM-AN017
COMPARE
0.130" 1
₹680.00INR / Each
Nordson Divert 3-Way Stopcock, Polycarbonate, Female Luer x Female Luer x 0.110 Bond Port; 25/Pk 5614093 Mfr # FCVFFM-030
COMPARE
0.110" 25
₹8,700.00INR / Pkg of 25
Nordson Divert 3-Way Stopcock, Polycarbonate, Female Luer x Female Luer x 0.130" Bond Port; 25/Pk 5614094 Mfr # FCVFFM-AN017
COMPARE
0.130" 25
₹9,700.00INR / Pkg of 25