Nordson Divert 3-Way Stopcock, Polycarbonate, Female Luer x Female Luer x 0.110" Bond Port; 1/Ea
Representative image only.
Nordson Divert 3-Way Stopcock, Polycarbonate, Female Luer x Female Luer x 0.110" Bond Port; 1/Ea
| Mfr: FCVFFM-030| Item: 5614085
Easily control the flow of a liquid or gas using an on/off diverter
₹600.00INR / Each
₹600.00INR / Each
Product Description
Key Features
  • Low-torque handle provides smooth adjustments
  • Diverter arrow indicates off position
  • DEHP free
  • ISO 80369-7 compliant
  • HPDE diverter
  • Female Luer x Female Luer x 0.110 Bond Port
  • Accommodates a 0.110" OD tube
  • 1 each
  • Manufacturer defect warranty
More About this Item
Add a stopcock to your system for an easy on/off control of your fluid. Three-way stopcocks feature two female Luer fittings plus a bond port. Luer fittings on stopcocks ensure a quick, leak-free connection; bond port creates a permanent connection, simply match the tube OD to bond port size. Diverter arrow indicates off position, enhancing operational efficiency and minimizing errors. Transparent polycarbonate body makes it easy to see the flow.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPolycarbonate
  • Number Of Ports3
  • Connection TypeFemale Luer x Female Luer x Bond Port
  • Max Operating Pressure (PSI)50
  • Regulatory ApprovalsNon-DEHP
  • Luer Connection Type (Port 1)Female Luer
  • Luer Connection Type (Port 2)Female Luer
  • Bond Port0.110"
  • Quantity1
  • Description3-Way Stopcock, Polycarbonate, Female Luer x Female Luer x 0.110 Bond Port; 1/Ea
  • WarrantyManufacturer defect