Nomex IIIA Flame-Resistant Lab Coats
Constructed of flame-resistant Nomex IIIA fabric
Nomex IIIA fabric is inherently flame-resistant — no need to worry about washing out the protection! Lab coats feature three outside pockets, side vent openings, hemmed sleeve ends, and a lapel collar. Rated to Arc Thermal Protective Value (ATPV) of 5.1 calories/cm2 and a Hazard Risk Category level of HRC 1. Meets the requirements of NFPA 2112, Standard on Flame-Resistant Garments for Protection of Industrial Personnel Against Flash Fire. Color is royal blue.
40,000.00 - ₹69,300.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Length (in) Size
Availability Pricing
VF Workwear Nomex IIIA Flame-Resistant Lab Coat, Small 8646850 Mfr # KNL2RB-SMALL (PACK BY PERSON)
COMPARE
Royal Blue 41.5 Small
₹69,300.00INR / Each
VF Workwear Nomex IIIA Flame-Resistant Lab Coat, Medium 8646851 Mfr # KNL2RB-MEDIUM (PACK BY PERSON)
COMPARE
Royal Blue 41.5 Medium
₹40,000.00INR / Each
VF Workwear Nomex IIIA Flame-Resistant Lab Coat, Large 8646852 Mfr # KNL2RB-LARGE (PACK BY PERSON)
COMPARE
Royal Blue 41.5 Large
₹40,300.00INR / Each
VF Workwear Nomex IIIA Flame-Resistant Lab Coat, X-large 8646853 Mfr # KNL2RB-XLARGE (PACK BY PERSON)
COMPARE
Royal Blue 41.5 X-large
₹40,300.00INR / Each