VF Workwear Nomex IIIA Flame-Resistant Lab Coat, Small
Representative image only.
VF Workwear Nomex IIIA Flame-Resistant Lab Coat, Small
VF Workwear | Mfr: KNL2RB-SMALL (PACK BY PERSON)| Item: 8646850
Constructed of flame-resistant Nomex IIIA fabric
₹69,300.00INR / Each
₹69,300.00INR / Each
Product Description
Key Features
  • 3 month warranty
More About this Item
Nomex IIIA fabric is inherently flame-resistant — no need to worry about washing out the protection! Lab coats feature three outside pockets, side vent openings, hemmed sleeve ends, and a lapel collar. Rated to Arc Thermal Protective Value (ATPV) of 5.1 calories/cm2 and a Hazard Risk Category level of HRC 1. Meets the requirements of NFPA 2112, Standard on Flame-Resistant Garments for Protection of Industrial Personnel Against Flash Fire. Color is royal blue.
3 Month Warranty
Specifications
  • Length (in)41.5
  • Length (cm)105.4
  • SizeSmall
  • ColorRoyal Blue
  • DescriptionNomex IIIA Flame-Resistant Lab Coat, Small