NFPA Right-To-Know Hazard Communication Labels
Easily identify the severity of the health hazard, flammability, instability, and the necessary personal protective equipment
  • Writable surface—perfect for writing in any necessary information
  • Self-adhesive and made of pressure sensitive paper
National Fire Protection Association (NFPA) labels convey general information to emergency personnel regarding the handling of hazardous chemicals. Easily identify the severity of the health hazard, flammability, instability, and the necessary personal protective equipment. Write in the information needed for your particular use. Labels are self-adhesive and made of pressure sensitive paper.
Compliance: OSHA 29 CFR1910.1200
3,800.00 - ₹8,100.00INR / Roll of 250
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Height (in) Width (in)
Availability Pricing
National Marker HMS12L Right-To-Know Hazard NFPA Label, 2" x 2"; 250/Roll 8639603 Mfr # HMS12L
COMPARE
2 2
₹3,800.00INR / Roll of 250
National Marker HMS14L Right-To-Know Hazard NFPA Label, 4" x 4"; 250/Roll 8639604 Mfr # HMS14L
COMPARE
4 4
₹8,100.00INR / Roll of 250
National Marker WML1 Right-To-Know Hazard NFPA Label, 7/8" x 7/8"; 750/Roll 8639611 Mfr # WML1
COMPARE
0.875 0.875
₹5,000.00INR / Roll of 750
National Marker WOL3 Right-To-Know Hazard NFPA Label, 2-7/8" x 7/8"; 500/Roll 8639613 Mfr # WOL3
COMPARE
0.875 2.875
₹6,600.00INR / Roll of 500