National Marker HMS14L Right-To-Know Hazard NFPA Label, 4" x 4"; 250/Roll

Representative image only.

National Marker HMS14L Right-To-Know Hazard NFPA Label, 4" x 4"; 250/Roll

National Marker | Mfr: HMS14L| Item: 8639604

Easily identify the severity of the health hazard, flammability, instability, and the necessary personal protective equipment

₹8,100.00INR / Roll of 250

₹8,100.00INR / Roll of 250
Product Description

Key Features

  • Writable surface—perfect for writing in any necessary information
  • Self-adhesive and made of pressure sensitive paper
  • Each label measures 4" x 4"

More About this Item

National Fire Protection Association (NFPA) labels convey general information to emergency personnel regarding the handling of hazardous chemicals. Easily identify the severity of the health hazard, flammability, instability, and the necessary personal protective equipment. Write in the information needed for your particular use. Labels are self-adhesive and made of pressure sensitive paper.
Compliance: OSHA 29 CFR1910.1200

Specifications
  • HeaderHAZARDOUS
  • LanguagesEnglish
  • MaterialPaper
  • ComplianceOSHA 29 CFR1910.1200
  • Width (in)4
  • Height (in)4
  • DescriptionRight-To-Know Hazard NFPA Label, 4" x 4"; 250/Roll
Reviews
Q&A