Nextteq® VeriAir Flex™ Air Sampling Bags
Easily collect air samples anywhere, at any time
  • Take an air sample without a pump Ideal for sampling in hot zones Lightweight, compact air sample bag can be easily stored and carried Handles made of static dissipative materials Stainless steel valve offers excellent chemical resistance
The VeriAir Flex sampling bags offer the convenience of collecting air samples without the use of a pump, saving you time and money. Simply open the valve, pull open the bag using the handles to let the air in, then close the valve. Air sample is ready to be tested with a portable gas chromatograph or other analytical instrument.
14,100.00 - ₹39,600.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Volume (Liters) Description
Availability Pricing
Nextteq NXT-31000 Manual Inflating Bag, VeriAir Flex Foil, 1 L 0800278 Mfr # NXT-31000
COMPARE
Foil 1 Manual Inflating Bag, VeriAir Flex Foil, 1 L
₹18,900.00INR / Each
Nextteq NXT-31029 Manual Inflating Bag, VeriAir Flex Foil, 5 L 0800279 Mfr # NXT-31029
COMPARE
Foil 5 Manual Inflating Bag, VeriAir Flex Foil, 5 L
₹22,100.00INR / Each
Nextteq NXT-90520 Manual Inflating Bag, VeriAir Flex Foil, 10 L 0800281 Mfr # NXT-90520
COMPARE
Foil 10 Manual Inflating Bag, VeriAir Flex Foil, 10 L
₹39,600.00INR / Each
Nextteq NXT-31019 Manual Inflating Bag, VeriAir Tedlar, 1 L 0800282 Mfr # NXT-31019
COMPARE
Tedlar 1 Manual Inflating Bag, VeriAir Tedlar, 1 L
₹14,100.00INR / Each