Nextteq NXT-90520 Manual Inflating Bag, VeriAir Flex Foil, 10 L
Representative image only.
Nextteq NXT-90520 Manual Inflating Bag, VeriAir Flex Foil, 10 L
| Mfr: NXT-90520| Item: 0800281
Easily collect air samples anywhere, at any time
₹39,600.00INR / Each
₹39,600.00INR / Each
Product Description
Key Features
  • Take an air sample without a pump Ideal for sampling in hot zones Lightweight, compact air sample bag can be easily stored and carried Handles made of static dissipative materials Stainless steel valve offers excellent chemical resistance
More About this Item
The VeriAir Flex sampling bags offer the convenience of collecting air samples without the use of a pump, saving you time and money. Simply open the valve, pull open the bag using the handles to let the air in, then close the valve. Air sample is ready to be tested with a portable gas chromatograph or other analytical instrument.
Specifications
  • MaterialFoil
  • Volume (Liters)10
  • DescriptionManual Inflating Bag, VeriAir Flex Foil, 10 L