NECi Nitrate Test Kits
Get quick, reliable non-toxic results
  • Eliminates the use of cadmium, a known carcinogen
  • Sensitive and accurate—no drift or inconsistency in testing results over time
  • Testing can be easily done in the field or laboratory
  • Enzyme-based nitrate test kits are environmentally friendly
Nitrate testing kits provide accurate and reliable results without the use of cadmium, a toxic heavy metal and known carcinogen. All kits use the naturally occurring enzyme called nitrate reductase which safely measures nitrate in any aqueous sample.
The laboratory nitrate analysis method has been validated and approved by ASTM International, USGS, and as an Alternate Test Procedure under the Safe Drinking Water Act with the US EPA.
9,800.00 - ₹104,000.00INR / Pkg of 5
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
Compareitem specifications
Measurement Points Description
Availability Pricing
NECi Colorimetric Nitrate Water Test Kit, Standard Range, 5/Pk WW-05543-20 Mfr # NTK-WSR-05
COMPARE
0.5 - 10.0 ppm Colorimetric Nitrate Water Test Kit, Standard Range, 5/Pk
₹9,800.00INR / Pkg of 5
NECi Colorimetric Nitrate Water Test Kit, Standard Range, 10/Pk WW-05543-21 Mfr # NTK-WSR-10
COMPARE
0.5 - 10.0 ppm Colorimetric Nitrate Water Test Kit, Standard Range, 10/Pk
₹24,800.00INR / Pkg of 10
NECi Colorimetric Nitrate Water Test Kit, Standard Range, 25/Pk WW-05543-22 Mfr # NTK-WSR-25
COMPARE
0.5 - 10.0 ppm Colorimetric Nitrate Water Test Kit, Standard Range, 25/Pk
₹55,400.00INR / Pkg of 25
NECi Colorimetric Nitrate Water Test Kit, Standard Range, 50/Pk WW-05543-23 Mfr # NTK-WSR-50
COMPARE
0.5 - 10.0 ppm Colorimetric Nitrate Water Test Kit, Standard Range, 50/Pk
₹73,300.00INR / Pkg of 50
NECi Colorimetric Nitrate Water Test Kit, Low Range, 5/Pk WW-05543-24 Mfr # NTK-WLR-05
COMPARE
0.05 - 2.0 ppm Colorimetric Nitrate Water Test Kit, Low Range, 5/Pk
₹13,800.00INR / Pkg of 5
NECi Colorimetric Nitrate Water Test Kit, Low Range, 10/Pk WW-05543-25 Mfr # NTK-WLR-10
COMPARE
0.05 - 2.0 ppm Colorimetric Nitrate Water Test Kit, Low Range, 10/Pk
₹17,600.00INR / Pkg of 10
NECi Colorimetric Nitrate Water Test Kit, Low Range, 25/Pk WW-05543-26 Mfr # NTK-WLR-25
COMPARE
0.05 - 2.0 ppm Colorimetric Nitrate Water Test Kit, Low Range, 25/Pk
₹39,100.00INR / Pkg of 25
NECi Colorimetric Nitrate Water Test Kit, Low Range, 50/Pk WW-05543-27 Mfr # NTK-WLR-50
COMPARE
0.05 - 2.0 ppm Colorimetric Nitrate Water Test Kit, Low Range, 50/Pk
₹73,300.00INR / Pkg of 50
NECi Colorimetric Nitrate Soil Test Kit, 5/Pk WW-05543-28 Mfr # NTK-S-05
COMPARE
0 - 50 ppm Colorimetric Nitrate Soil Test Kit, 5/Pk
₹9,800.00INR / Pkg of 5
NECi Colorimetric Nitrate Soil Test Kit, 10/Pk WW-05543-29 Mfr # NTK-S-10
COMPARE
0 - 50 ppm Colorimetric Nitrate Soil Test Kit, 10/Pk
₹24,800.00INR / Pkg of 10
NECi Colorimetric Nitrate Soil Test Kit, 25/Pk WW-05543-30 Mfr # NTK-S-25
COMPARE
0 - 50 ppm Colorimetric Nitrate Soil Test Kit, 25/Pk
₹55,400.00INR / Pkg of 25
NECi Colorimetric Nitrate Soil Test Kit, 50/Pk WW-05543-31 Mfr # NTK-S-50
COMPARE
0 - 50 ppm Colorimetric Nitrate Soil Test Kit, 50/Pk
₹104,000.00INR / Pkg of 50
NECi Test Tube Nitrate Water Test Kit, Standard Range, 25/Pk WW-05543-35 Mfr # NTK-TTSR-25
COMPARE
1.0 - 10.0 ppm Test Tube Nitrate Water Test Kit, Standard Range, 25/Pk
₹12,200.00INR / Pkg of 25
NECi Test Tube Nitrate Water Test Kit, Low Range, 25/Pk WW-05543-36 Mfr # NTK-TTLR-25
COMPARE
0.05 - 1.0 ppm Test Tube Nitrate Water Test Kit, Low Range, 25/Pk
₹12,200.00INR / Pkg of 25
NECi Test Tube Nitrate Water Test Kit, Standard Range, 100/Pk WW-05543-37 Mfr # NTK-TTSR-100
COMPARE
1.0 - 10.0 ppm Test Tube Nitrate Water Test Kit, Standard Range, 100/Pk
₹41,500.00INR / Pkg of 100
NECi Test Tube Nitrate Water Test Kit, Low Range, 100/Pk WW-05543-38 Mfr # NTK-TTLR-100
COMPARE
0.05 - 1.0 ppm Test Tube Nitrate Water Test Kit, Low Range, 100/Pk
₹30,500.00INR / Pkg of 100
NECi Test Reagent Kit for Tabletop Discreet Analyzers WW-05543-39 Mfr # NRPK-DA-1AT
COMPARE
Test Reagent Kit for Tabletop Discreet Analyzers
₹12,100.00INR / Pkg
NECi Test Reagent Kit for High Capacity Discreet Analyzers WW-05543-40 Mfr # NRPK-HDA-3AT
COMPARE
Test Reagent Kit for High Capacity Discreet Analyzers
₹28,500.00INR / Pkg
NECi Test Reagent Kit for Flow Injection Analyzers WW-05543-41 Mfr # NRPK-FIA-3AT
COMPARE
Test Reagent Kit for Flow Injection Analyzers
₹24,400.00INR / Pkg
Showing 1- 19 of 19